Skip to Content
Merck
All Photos(4)

Key Documents

HPA014736

Sigma-Aldrich

Anti-SLC13A3 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Na(+)/dicarboxylate cotransporter 3, Anti-NaDC-3, Anti-Sodium-dependent high-affinity dicarboxylate transporter 2, Anti-Solute carrier family 13 member 3, Anti-hNaDC3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

SLFGQKEVRKDPSQESEENTAAVRRNGLHTVPTEMQFLASTEAKDHPGETEVPLDLPADSRKEDEYRRNIWKGFL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC13A3(64849)

General description

SLC13A3 (solute carrier family 13, member 3) belongs to SLC13 family of five members. This family is divided into Na+-sulphate (NaS) cotransporters and Na+-carboxylate (NaC) cotransporters, and SLC13A3 belongs to the latter group. It is a transporter with 12 transmembrane regions, and is composed of 602 amino acids. This gene maps to human chromosome 20q12-13.1, spans more than 80kb, and has 13 exons, and 12 introns. It has three predicted N-linked glycosylation sites. It is localized to the proximal tubule cells in the basolateral membrane. It is also expressed in brain, liver, pancreas and placenta.

Immunogen

Solute carrier family 13 member 3 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

SLC13A3 (solute carrier family 13, member 3) acts in collaboration with organic anion transporter 1 (OAT1) and OAT3) to facilitate the release of multiple endogenous and exogenous organic ions. It provides intracellular α-ketoglutarate, in a Na+-dependent manner, to OAT1 and 3, to allow for organic anion/dicarboxylate exchange. It is a glutathione (GSH) transporter of low-affinity. It is responsible for the uptake of GSH by the proximal tubule cells of basolateral membrane, in a sodium-dependent manner. Mutations in this gene might have a link to idiopathic nephrolithiasis. This transporter is also responsible for the uptake of N-carbamoylglutamate (NCG) from blood. NCG is a drug which is used to treat inborn n-acetylglutamate synthase deficiency. SLC13A3 might be involved in myelination, as it is responsible for the Na+-dependent transport of N-acetylaspartate in brain.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73142

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lena Schorbach et al.
Nephron. Physiology, 124(1-2), 1-5 (2013-11-20)
During a single pass through the kidneys, more than 80% of glutathione (GSH) is excreted, indicating not only glomerular filtration, but also tubular secretion. The first step in tubular secretion is the uptake of a substance across the basolateral membrane
W Huang et al.
The Journal of pharmacology and experimental therapeutics, 295(1), 392-403 (2000-09-19)
N-Acetylaspartate is a highly specific marker for neurons and is present at high concentrations in the central nervous system. It is not present at detectable levels anywhere else in the body other than brain. Glial cells express a high-affinity transporter
Daniel Markovich et al.
Pflugers Archiv : European journal of physiology, 447(5), 594-602 (2003-08-14)
The SLC13 gene family consist of five sequence-related members that have been identified in a variety of animals, plants, yeast and bacteria. Proteins encoded by these genes are divided into two functionally unrelated groups: the Na(+)-sulphate (NaS) cotransporters and the
Elisabeth Schwob et al.
American journal of physiology. Renal physiology, 307(12), F1373-F1379 (2014-10-31)
Inborn defects in N-acetylglutamate (NAG) synthase (NAGS) cause a reduction of NAG, an essential cofactor for the initiation of the urea cycle. As a consequence, blood ammonium concentrations are elevated, leading to severe neurological disorders. The orphan drug N-carbamoylglutamate (NCG;
H Wang et al.
American journal of physiology. Cell physiology, 278(5), C1019-C1030 (2000-05-04)
We have cloned and functionally characterized the human Na(+)-dependent high-affinity dicarboxylate transporter (hNaDC3) from placenta. The hNaDC3 cDNA codes for a protein of 602 amino acids with 12 transmembrane domains. When expressed in mammalian cells, the cloned transporter mediates the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service