Skip to Content
Merck
All Photos(7)

Key Documents

HPA012624

Sigma-Aldrich

Anti-RHOT2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-MIRO-2, Anti-Mitochondrial Rho GTPase 2, Anti-Ras homolog gene family member T2, Anti-hMiro-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
conjugate:
unconjugated
application:
IF
IHC
clone:
polyclonal
species reactivity:
human
citations:
5
technique(s):
immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

RSCLGHLGYLGYPTLCEQDQAHAITVTREKRLDQEKGQTQRSVLLCKVVGACGVGKSAFLQAFLGRGLGHQDTREQPPGYAIDTVQVNGQEKYLILC

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RHOT2(89941)

General description

Mitochondrial Rho GTPase 2 (RHOT2) is a GTPase having a C-terminal transmembrane domain which targets it to the mitochondria. It also has GTP-binding domains which are separated by a linker region with calcium-binding motifs.

Immunogen

Mitochondrial Rho GTPase 2 recombinant protein epitope signature tag (PrEST)

Application

Anti-RHOT2 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting at a dilution of 1:500. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

Mitochondrial Rho GTPase 2 (RHOT2) is involved in mediating mitochondrial transport. It interacts with hypoxia up-regulated mitochondrial movement regulator (HUMMR) which influences mitochondrial transport in the anterograde direction. RHOT2 induces the aggregation of thread-like mitochondria but doesn′t take part in their formation. Along with Mitofusin 2 (Mfn2), it takes part in mediating axonal mitochondrial transport and also links mitochondria to kinesin.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72007

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicol Birsa et al.
The Journal of biological chemistry, 289(21), 14569-14582 (2014-03-29)
Mitochondrial transport plays an important role in matching mitochondrial distribution to localized energy production and calcium buffering requirements. Here, we demonstrate that Miro1, an outer mitochondrial membrane (OMM) protein crucial for the regulation of mitochondrial trafficking and distribution, is a
Albert Misko et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 30(12), 4232-4240 (2010-03-26)
Mitofusins (Mfn1 and Mfn2) are outer mitochondrial membrane proteins involved in regulating mitochondrial dynamics. Mutations in Mfn2 cause Charcot-Marie-Tooth disease (CMT) type 2A, an inherited disease characterized by degeneration of long peripheral axons, but the nature of this tissue selectivity
Yan Li et al.
The Journal of cell biology, 185(6), 1065-1081 (2009-06-17)
Mitochondrial transport is critical for maintenance of normal neuronal function. Here, we identify a novel mitochondria protein, hypoxia up-regulated mitochondrial movement regulator (HUMMR), which is expressed in neurons and is markedly induced by hypoxia-inducible factor 1 alpha (HIF-1alpha). Interestingly, HUMMR
Sa Fransson et al.
Biochemical and biophysical research communications, 344(2), 500-510 (2006-04-25)
We recently described the atypical Rho GTPases Miro-1 and Miro-2. These proteins have tandem GTP-binding domains separated by a linker region with putative calcium-binding motives. In addition, the Miro GTPases have a C-terminal transmembrane domain, which confers targeting to the
Chung-Han Hsieh et al.
Cell metabolism, 30(6), 1131-1140 (2019-10-01)
The identification of molecular targets and pharmacodynamic markers for Parkinson's disease (PD) will empower more effective clinical management and experimental therapies. Miro1 is localized on the mitochondrial surface and mediates mitochondrial motility. Miro1 is removed from depolarized mitochondria to facilitate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service