Skip to Content
Merck
All Photos(9)

Key Documents

HPA011954

Sigma-Aldrich

Anti-ASGR1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-ASGP-R 1, Anti-ASGPR 1, Anti-Asialoglycoprotein receptor 1, Anti-Hepatic lectin H1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
recombinant expression
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

MTKEYQDLQHLDNEESDHHQLRKGPPPPQPLLQRLCSGPRL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ASGR1(432)

General description

ASGR1 (Asialoglycoprotein receptor 1) is a Ca2+-dependent, hepatocyte-specific, lectin membrane protein involved in hepatitis B virus (HBV) binding and uptake. This 40-50kDa protein is expressed at the sinusoidal and basolateral hepatocellular membrane of the human liver. It is a hetero-oligomeric compound composed of two homologous, covalently bound polypeptides, called HL-1 (Hepatic Lectin, or ASGPR1, ASGR1) and HL-2 (ASGPR2, ASGR2), with HL-1 as the major subunit.

Immunogen

Asialoglycoprotein receptor 1 recombinant protein epitope signature tag (PrEST)

Application

Anti-ASGR1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

The transmembrane protein, ASGR1 (Asialoglycoprotein receptor 1), is highly involved in the endocytosis and lysosomal degradation of desialylated proteins from the serum. It binds to desialylated proteins and degrades the extracellular glycoproteins with the release of terminal galactose, lactose or N-acetyl-galactosamine residues. It has been reported that ASGR1 expression is upregulated in embryonic stem cells upon hepatic differentiation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72012

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dan Xu et al.
Cell transplantation, 23(12), 1573-1584 (2013-10-24)
We developed a novel method for differentiating adipocyte-derived stem cells (ASCs) into hepatocyte-like cells (iHeps). ASCs are cultured as spherical cellular aggregates and are then induced by culture in chemically defined media for a short time period to differentiate into
Dominik Witzigmann et al.
Hepatology research : the official journal of the Japan Society of Hepatology, 46(7), 686-696 (2015-10-01)
One of the most promising strategies for the treatment of liver diseases is targeted drug delivery via the asialoglycoprotein receptor (ASGPR). The success of this approach heavily depends on the ASGPR expression level on parenchymal liver cells. In this study
Massimiliano Paganelli et al.
Hepatology (Baltimore, Md.), 57(1), 59-69 (2012-08-18)
The role of cell differentiation state on hepatitis B virus (HBV) replication has been well demonstrated, whereas how it determines cell susceptibility to HBV entry is far less understood. We previously showed that umbilical cord matrix stem cells (UCMSC) can
Ashish Kumar Vyas et al.
Liver international : official journal of the International Association for the Study of the Liver, 38(12), 2149-2158 (2018-05-02)
Asialoglycoprotein receptor expression on hepatocytes has been associated with endocytosis, binding and uptake of hepatitis B virus. The role of asialoglycoprotein receptor in hepatitis B virus vertical transmission and its expression on placenta has not yet been studied. Thirty-four HBsAg+ve
Bin Shi et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 61(12), 901-909 (2013-08-28)
Human hepatocellular carcinoma (HCC) is the fifth most common cancer in the world. Currently, surgical resection is the only effective treatment for HCC if the tumor is resectable. Small molecule, biologics and siRNA anti-cancer drugs have been explored for the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service