Skip to Content
Merck
All Photos(2)

Key Documents

HPA005968

Sigma-Aldrich

Anti-HLX antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-HLX1, Anti-Homeobox protein HB24, Anti-Homeobox protein HLX1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

CSFPLDPAAVKKPSFCIADILHAGVGDLGAAPEGLAGASAAALTAHLGSVHPHASFQAAARSPLRPTPVVAPSEVPAGFPQRLSPLPAAYHHHHPQQQQQQQ

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HLX(3142)

Immunogen

H2.0-like homeobox protein recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

H2.0-like homeobox protein is a protein encoded by the HLX gene in humans. It is referred as HB24 and HLX1. It is expressed in proliferating and migrating human trophoblast cells. Its expression is found to decreased significantly in human FGR. It regulates human trophoblast-derived cell migration, and that hepatocyte growth factor (HGF), by receptor c-met, acts through this gene to control cell migration. Polymorphisms in the this gene increase the risk for childhood asthma. It is primarily expressed in cytotrophoblast cell types in the human placenta and is involved in cytotrophoblast proliferation and down-regulation of cell differentiation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST84766

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gayathri Rajaraman et al.
Reproduction, fertility, and development, 20(3), 357-367 (2008-04-12)
Homeobox genes are a large family of transcription factors. Of these, the HLX homeobox gene (previously known as HLX1 and HB24) is important for normal placentation. We have previously shown that HLX mRNA expression is significantly reduced in fetal growth-restricted
Kathrin Suttner et al.
The Journal of allergy and clinical immunology, 123(1), 82-88 (2008-11-29)
Major transcription factors controlling T(H)1 and T(H)2 development, such as T-box transcription factor and GATA3, might be centrally involved in asthma and atopic diseases. Only recently, the homeobox transcription factor H.20-like homeobox 1 (HLX1), interacting closely with T-box transcription factor
Charles Y Lin et al.
Nature, 530(7588), 57-62 (2016-01-28)
Medulloblastoma is a highly malignant paediatric brain tumour, often inflicting devastating consequences on the developing child. Genomic studies have revealed four distinct molecular subgroups with divergent biology and clinical behaviour. An understanding of the regulatory circuitry governing the transcriptional landscapes
Gayathri Rajaraman et al.
Biology of reproduction, 83(4), 676-683 (2010-06-18)
Homeobox gene transcription factors play a critical role in normal placental development and are expressed in specialized trophoblast cells. Abnormal trophoblast function is associated with clinically significant pregnancy disorders, including fetal growth restriction (FGR). Our previous studies demonstrated that homeobox

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service