Skip to Content
Merck
All Photos(1)

Key Documents

HPA005471

Sigma-Aldrich

Anti-PSMC4 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-26S protease regulatory subunit 6B antibody produced in rabbit, Anti-MB67-interacting protein antibody produced in rabbit, Anti-MIP224 antibody produced in rabbit, Anti-Proteasome 26S subunit ATPase 4 antibody produced in rabbit, Anti-TAT-binding protein 7 antibody produced in rabbit, Anti-TBP-7 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PSMC4(5704)

Looking for similar products? Visit Product Comparison Guide

General description

The 26S proteasome is an ATP-dependent protease, which is a large complex of multiple subunits. It contains one catalytic subunit, 20S and two regulatory subunits called PA700. In humans, the PA700 contains six members, incluing PSMC4 (proteasome 26S subunit, ATPase, 4). It is also called TBP7 and the corresponding gene is located on chromosome 19q13.11-q13.13. PSMC4 is an ATPase and was initially identified as synphilin-1-interacting protein. Its molecular weight is around 50kDa.

Immunogen

26S protease regulatory subunit 6B recombinant protein epitope signature tag (PrEST)

Sequence
LTSDQKPDVMYADIGGMDIQKQEVREAVELPLTHFELYKQIGIDPPRGVLMYGPPGCGKTMLAKAVAHHTTAAFIRVVGSEFVQKYLGEGPRMVRDVFRLAKENAPAIIFIDEIDAIATKRFDAQTGADREVQRILLEL

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Proteasome 26S subunit, ATPase, 4 (PSMC4) regulates the assembly of the 26S proteasome complex. The HbYX motif present at the C-terminal of PSMC4 is essential for the assembly of 26S complex from PA700 and 20S subunits. Interaction of PSMC4 and TRAP1 (Tumor necrosis factor receptor-associated protein-1) leads to increased stress-induced cell death and intracellular protein ubiquitination. Studies also suggest that PSMC4, along with TRAP1, regulates the quality of mitochondria targeted proteins. An insertion/deletion mutation in intron 5 of this gene is found in higher frequency in Parkinson′s disease patients than in control. This mutation is less frequent in late-onset PD patients as opposed to early onset PD patients.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86558

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M R Amoroso et al.
Cell death and differentiation, 19(4), 592-604 (2011-10-08)
Tumor necrosis factor receptor-associated protein-1 (TRAP1) is a mitochondrial (MITO) antiapoptotic heat-shock protein. The information available on the TRAP1 pathway describes just a few well-characterized functions of this protein in mitochondria. However, our group's use of mass-spectrometric analysis identified TBP7
Brajesh Kumar et al.
The Journal of biological chemistry, 285(50), 39523-39535 (2010-10-13)
PA700, the 19 S regulatory subcomplex of the 26 S proteasome, contains a heterohexameric ring of AAA subunits (Rpt1 to -6) that forms the binding interface with a heteroheptameric ring of α subunits (α1 to -7) of the 20 S
Claudia Wahl et al.
Journal of neural transmission (Vienna, Austria : 1996), 115(8), 1141-1148 (2008-05-01)
Dysfunction of proteasomal protein degradation is involved in neurodegeneration in Parkinson's disease (PD). Recently we identified the regulatory proteasomal subunit S6 ATPase as a novel interactor of synphilin-1, which is a substrate of the ubiquitin-ligase Parkin (PARK2) and an interacting
N Tanahashi et al.
Biochemical and biophysical research communications, 243(1), 229-232 (1998-02-25)
The 26S proteasome is a eukaryotic ATP-dependent protease functioning as a protein death machine. It is a large multisubunit complex, consisting of a catalytic 20S proteasome and two regulatory modules, named PA700. The PA700 complex is composed of multiple subunits

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service