Skip to Content
Merck
All Photos(4)

Key Documents

HPA004883

Sigma-Aldrich

Anti-MYEF2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-MST156 antibody produced in rabbit, Anti-MyEF-2 antibody produced in rabbit, Anti-Myelin expression factor 2 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

GMGSMNSVTGGMGMGLDRMSSSFDRMGPGIGAILERSIDMDRGFLSGPMGSGMRERIGSKGNQIFVRNLPFDLTWQKLKEKFSQCGHVMFAEIKMENGKSKGCGTVRFDSPESAEKACRIMNGIKISGREIDVRLDRN

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MYEF2(50804)

Related Categories

General description

MYEF2 (myelin expression factor 2) is a member of the myocyte enhancer factor-2 (MEF2) family of MADS (MCM1, agamous, deficiens, serum response factor)-box transcription factors. It reflects a strong response to the cell division, differentiation, and cell death in a calcium-dependent manner.

Immunogen

Myelin expression factor 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

MYEF2 (myelin expression factor 2) acts as a repressor protein of the myelin basic protein (MBP). MBP is the main component of the myelin sheath. During erythropoiesis, it represses hematopoietic genes in erythroid cells with the help of transcription factor RUNX1. It is involved in MBP associated transcriptional regulation during the brain development. It is highly involved in myogenesis and morphogenesis. It also regulates cell proliferation during the development of skeletal, cardiac, and visceral muscle through several growth factor-regulated intracellular signaling pathways.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86921

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B L Black et al.
Annual review of cell and developmental biology, 14, 167-196 (1999-01-19)
Metazoans contain multiple types of muscle cells that share several common properties, including contractility, excitability, and expression of overlapping sets of muscle structural genes that mediate these functions. Recent biochemical and genetic studies have demonstrated that members of the myocyte
Timothy A McKinsey et al.
Trends in biochemical sciences, 27(1), 40-47 (2002-02-14)
The decision of a cell to divide, differentiate or die is dependent on the coupling of cytoplasmic signals to the activation and repression of specific sets of genes in the nucleus. Many of the signal transduction pathways that control these
V Muralidharan et al.
Journal of cellular biochemistry, 66(4), 524-531 (1997-09-15)
Myelin basic protein (MBP) is a major component of the myelin sheath whose production is developmentally controlled during myelinogenesis. Earlier studies have indicated that programmed expression of the MBP gene is regulated at the level of transcription. Evidently, the MB1
Boet van Riel et al.
Molecular and cellular biology, 32(19), 3814-3822 (2012-07-18)
RUNX1 is known to be an essential transcription factor for generating hematopoietic stem cells (HSC), but much less is known about its role in the downstream process of hematopoietic differentiation. RUNX1 has been shown to be part of a large

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service