Skip to Content
Merck
All Photos(1)

Key Documents

H7768

Sigma-Aldrich

12(S)-Hydroxy-(5Z,8Z,10E,14Z)-eicosatetraenoic acid

~100 μg/mL in ethanol, ≥95% (HPLC)

Synonym(s):

(S)-12-HETE

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H32O3
CAS Number:
Molecular Weight:
320.47
Beilstein:
2656104
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

liquid

concentration

~100 μg/mL in ethanol

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCC\C=C/C[C@H](O)\C=C\C=C/C\C=C/CCCC(O)=O

InChI

1S/C20H32O3/c1-2-3-4-5-10-13-16-19(21)17-14-11-8-6-7-9-12-15-18-20(22)23/h7-11,13-14,17,19,21H,2-6,12,15-16,18H2,1H3,(H,22,23)/b9-7-,11-8-,13-10-,17-14+/t19-/m0/s1

InChI key

ZNHVWPKMFKADKW-LQWMCKPYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

12(S)-HETE is a bioactive lipid implicated in angiogenesis, growth, and metastasic action in tumor cell lines and in animal models. Arachidonate 12-lipoxygenase (12-LOX) converts arachidonic acid to 12(S)-(HETE). Alteration in 12-LOX expression or activity has been reported in various carcinomas including prostate carcinoma.
Prometastatic metabolite of arachidonic acid via the 12-lipoxygenase pathway that is expressed in many tumor cell lines. It is produced in coronary microvasculature and is a potent inducer of relaxation and hyperpolarization of these microvessels. It activates large-conductance Ca2+-activated K+ currents in coronary microvascular smooth muscle.

Packaging

Packaged under Argon.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F

Flash Point(C)

13 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuke Li et al.
Free radical biology & medicine, 134, 239-247 (2019-01-20)
This study was designed to investigate the neuroprotective effects of baicalein and the effect of the cortical 12/15-lipoxygenase (12/15-LOX) pathway on diabetic cognitive dysfunction. Our results showed that spatial learning and memory ability, as well as cortex neurons, were significantly
Yan Qu et al.
Cancer management and research, 11, 8303-8313 (2019-10-02)
To clarify the effect of 12-lipoxygenase/12-hydroxyeicosatetraeonic acid (12-LOX/12-HETE) on progress of esophageal squamous cell carcinoma (ESCC) and the possible mechanism. We performed cell experiments including chemical treatment, transfection, Western blotting and transwell assay to investigate the function of 12-LOX/12-HETE. Slices
Alicja Pakiet et al.
Lipids in health and disease, 18(1), 29-29 (2019-01-28)
Altered metabolism of lipids is currently considered a hallmark characteristic of many malignancies, including colorectal cancer (CRC). Lipids are a large group of metabolites that differ in terms of their fatty acid composition. This review summarizes recent evidence, documenting many
C Vonach et al.
British journal of cancer, 105(2), 263-271 (2011-06-02)
The intravasation of breast cancer into the lymphendothelium is an early step of metastasis. Little is known about the mechanisms of bulky cancer invasion into lymph ducts. To particularly address this issue, we developed a 3-dimensional co-culture model involving MCF-7
Bei Wang et al.
Signal transduction and targeted therapy, 6(1), 94-94 (2021-02-28)
The arachidonic acid (AA) pathway plays a key role in cardiovascular biology, carcinogenesis, and many inflammatory diseases, such as asthma, arthritis, etc. Esterified AA on the inner surface of the cell membrane is hydrolyzed to its free form by phospholipase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service