Skip to Content
Merck
All Photos(1)

Key Documents

H0895

Sigma-Aldrich

Heparin disaccharide IV-A sodium salt

Synonym(s):

α-ΔUA-[1→4]-GlcNAc

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C14H20NO11Na
CAS Number:
Molecular Weight:
401.30
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

form

powder

Quality Level

color

white

storage temp.

−20°C

SMILES string

[Na].CC(=O)NC(C=O)C(O)C(OC1OC(=CC(O)C1O)C(O)=O)C(O)CO

InChI

1S/C14H21NO11.Na.H/c1-5(18)15-6(3-16)10(21)12(8(20)4-17)26-14-11(22)7(19)2-9(25-14)13(23)24;;/h2-3,6-8,10-12,14,17,19-22H,4H2,1H3,(H,15,18)(H,23,24);;

InChI key

KSLVSLXDGFKPRA-UHFFFAOYSA-N

Biochem/physiol Actions

Products of the digestion of heparin and heparan sulfate by various heparinases

Other Notes

In the following listings, ΔUA = 4-deoxy-L-threo-hex-4-enopyranosyluronic acid; GlcN = D-glucosamine; Ac = Acetyl; NS, 2S, 6S, = N-sulfo, 2-sulfate and 6-sulfate respectively.
To gain a comprehensive understanding of our extensive range of Disaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Preparation Note

Produced by the action of heparinase II and III

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Glycosaminoglycans are large linear polysaccharides constructed of repeating disaccharide units.

Glycosaminoglycans are large linear polysaccharides constructed of repeating disaccharide units.

Glycosaminoglycans are large linear polysaccharides constructed of repeating disaccharide units.

Glycosaminoglycans are large linear polysaccharides constructed of repeating disaccharide units.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service