Skip to Content
Merck
All Photos(1)

Key Documents

G113

Sigma-Aldrich

GR 64349

>96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H68N10O11S
CAS Number:
Molecular Weight:
921.11
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

>96%

storage temp.

−20°C

SMILES string

CSCC[C@H](NC(=O)[C@H](CC(C)C)N1CC[C@H](NC(=O)[C@@H](NC(=O)[C@H](Cc2ccccc2)NC(=O)[C@H](CO)NC(=O)[C@H](CC(O)=O)NC(=O)[C@@H](N)CCCCN)C(C)C)C1=O)C(N)=O

InChI

1S/C42H68N10O11S/c1-23(2)19-32(40(61)46-27(35(45)56)15-18-64-5)52-17-14-28(42(52)63)47-41(62)34(24(3)4)51-38(59)29(20-25-11-7-6-8-12-25)49-39(60)31(22-53)50-37(58)30(21-33(54)55)48-36(57)26(44)13-9-10-16-43/h6-8,11-12,23-24,26-32,34,53H,9-10,13-22,43-44H2,1-5H3,(H2,45,56)(H,46,61)(H,47,62)(H,48,57)(H,49,60)(H,50,58)(H,51,59)(H,54,55)/t26-,27-,28-,29-,30-,31-,32-,34-/m0/s1

InChI key

HVUNRXRFMQDMBO-NOJSRKIXSA-N

Gene Information

human ... TACR2(6865)
mouse ... TACR2(21337)
rat ... TACR2(25007)

Amino Acid Sequence

Lys-Asp-Ser-Phe-Val-Gly-R-γ-lactam-Leu-Met-NH2

General description

Neurokinin (NK) receptors are acted upon by tachykinins or neurokinins namely, NKA and NKB. These are endogenous ligands. An exogenous agonist/ligand for NK2 is GR 64349 peptide.

Biochem/physiol Actions

GR 64349 peptide acts as an agonist of neurokinin (NK2) receptor, which leads to reduced NK2 response to kainate and AMPA (α-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid), but not to NMDA (N-methyl-D-aspartate).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rajesh Gour et al.
European journal of medicinal chemistry, 178, 552-570 (2019-06-20)
We report herein the synthesis and anticancer activity of a set of novel S-linked artemisinins bearing an aliphatic/aromatic/heterocyclic nucleus as a substituent on the sulfur. The compounds were prepared from artemisinin via its lactol-form by an acid-catalyzed condensation of the
Lesley Marson et al.
European journal of pharmacology, 819, 261-269 (2017-12-15)
The suitability of various neurokinin-2 (NK2) receptor agonists and routes of administration to elicit on-demand voiding of the bladder and bowel, as future therapy for individuals with spinal cord injury, was examined using a rat model. The current study examined
S Guard et al.
Neurochemistry international, 18(2), 149-165 (1991-01-01)
The use of selective agonists in both functional and binding studies has provided unequivocal evidence for the existence of three types of tachykinin receptor (NK(1), NK(2) and NK(3)); there is also preliminary evidence for the existence of further subtypes. These
R M Hagan et al.
Neuropeptides, 19(2), 127-135 (1991-06-01)
The pharmacological profiles of two novel neurokinin agonists have been investigated. delta Ava[L-Pro9,N-MeLeu10]SP(7-11) (GR73632) and [Lys3,Gly8-R-gamma-lactam-Leu9] NKA(3-10) (GR64349) are potent and selective agonists at NK-1 and NK-2 receptors respectively. In the guinea-pig isolated trachea preparation, contractions induced by these agonists
B A Chizh et al.
British journal of pharmacology, 115(6), 1013-1019 (1995-07-01)
1. The effects of selective tachykinin (neurokinin, NK) NK1 and NK2 receptor antagonists have been examined on spinal neurones in alpha-chloralose anaesthetized, spinalized rats. They were tested for effects on responses both to excitatory amino acids (EAA) and to noxious

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service