EP0030128818
Eppendorf® Twin tec PCR plates, 384 wells
yellow plate, conical bottom black wells, pkg of 25 ea
All Photos(2)
About This Item
UNSPSC Code:
41106300
NACRES:
NB.15
material:
conical bottom black wells
yellow plate
yellow plate
size:
384 wells
feature:
skirt (Conical bottom)
Recommended Products
material
conical bottom black wells
yellow plate
feature
skirt (Conical bottom)
packaging
pkg of 25 ea
manufacturer/tradename
Eppendorf® 0030128818
technique(s)
PCR: suitable
size
384 wells
General description
“Eppendorf” Twin tec PCR plates with 384 wells are large scale PCR plates used in high throughput PCR workflows.
Application
Eppendorf “twin.tec” PCR Plates may be used in a wide variety of applications such as:
- Standard PCR
- Real time PCR
- High throughput PCR
Features and Benefits
- [TM=“Eppendorf”] Twin tec PCR plates have a unique one - piece design consisting of polypropylene wells and a rigid polycarbonate framework.
- Twin tec PCR plates have thin-walled polypropylene wells that ensure optimum heat transfer to the sample.
- The exceptionally solid polycarbonate framework is torque resistant.
- [TM=“Eppendorf”] Twin tec PCR plates show an improved well – well tolerance.
- The presence of raised well rims ensures effective sealing.
- [TM=“Eppendorf”] Twin tec PCR plates have been batch tested and certified DNase, RNase and PCR inhibitor free.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)