Skip to Content
Merck
All Photos(1)

Key Documents

EHU119551

Sigma-Aldrich

MISSION® esiRNA

targeting human SOCS3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CTTCAGCTCCAAGAGCGAGTACCAGCTGGTGGTGAACGCAGTGCGCAAGCTGCAGGAGAGCGGCTTCTACTGGAGCGCAGTGACCGGCGGCGAGGCGAACCTGCTGCTCAGTGCCGAGCCCGCCGGCACCTTTCTGATCCGCGACAGCTCGGACCAGCGCCACTTCTTCACGCTCAGCGTCAAGACCCAGTCTGGGACCAAGAACCTGCGCATCCAGTGTGAGGGGGGCAGCTTCTCTCTGCAGAGCGATCCCCGGAGCACGCAGCCCGTGCCCCGCTTCGACTGCGTGCTCAAGCTGGTGCACCACTACATGCCGCCCCCTGGAGCCCCCTCCTTCCCCTCGCCACCTACTGAACCCTCCTCCGAGGTGCCCGAGCAGCCGTCTGCCCAGCCACTCCCTGGGAGTCCCCCCAGAAGAGCCTATTACATCTACTCCGGGGGCGAGAAGATCCCCCTGGTGTTGAGCCGGCCCCTCTCCTCCAACGTGGCCACTCTTCAGCATCTCTGTCGGAAGACCGT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rak-Kyun Seong et al.
Pathogens (Basel, Switzerland), 9(3) (2020-03-04)
Zika virus (ZIKV) is a mosquito-borne flavivirus that has emerged and caused global outbreaks since 2007. Although ZIKV proteins have been shown to suppress early anti-viral innate immune responses, little is known about the exact mechanisms. This study demonstrates that
Yan Zhang et al.
Journal of neuroinflammation, 14(1), 211-211 (2017-11-04)
Morphine tolerance is a clinical challenge, and its pathogenesis is closely related to the neuroinflammation mediated by Toll-like receptor 4 (TLR4). In Chinese pain clinic, lidocaine is combined with morphine to treat chronic pain. We found that lidocaine sufficiently inhibited
Mandy Boontanrart et al.
Journal of neuroimmunology, 292, 126-136 (2016-03-06)
Microglia become activated immune cells during infection or disease in the central nervous system (CNS). However, the mechanisms that downregulate activated microglia to prevent immune-mediated damage are not completely understood. Vitamin D3 has been suggested to have immunomodulatory affects, and
Yujie Luo et al.
Stroke, 51(11), 3320-3331 (2020-09-17)
Neuroinflammation has been proven to play an important role in the pathogenesis of early brain injury after subarachnoid hemorrhage (SAH). EZH2 (enhancer of zeste homolog 2)-mediated H3K27Me3 (trimethylation of histone 3 lysine 27) has been recognized to play a critical
Naotoshi Iwahara et al.
Journal of Alzheimer's disease : JAD, 55(3), 1235-1247 (2016-11-05)
In response to changes of the central nervous system environment, microglia are capable of acquiring diverse phenotypes for cytotoxic or immune regulation and resolution of injury. Alzheimer's disease (AD) pathology also induces several microglial activations, resulting in production of pro-inflammatory

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service