Skip to Content
Merck

D2384

DL-3,4-Dihydroxyphenylserine

≥98% (HPLC), powder

Synonym(s):

DL-DOPS

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C9H11NO5
CAS Number:
Molecular Weight:
213.19
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.32
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to brown

solubility

0.1 M HCl: soluble (Solutions should be freshly prepared.), H2O: moderately soluble (Solutions should be freshly prepared.), aqueous base: unstable, ethanol: soluble (Solutions should be freshly prepared.)

storage temp.

−20°C

SMILES string

N[C@@H](C(O)c1cc(c(cc1)O)O)C(=O)O

InChI

1S/C9H11NO5/c10-7(9(14)15)8(13)4-1-2-5(11)6(12)3-4/h1-3,7-8,11-13H,10H2,(H,14,15)/t7-,8?/m0/s1

InChI key

QXWYKJLNLSIPIN-JAMMHHFISA-N

Biochem/physiol Actions

Amino acid precursor of noradrenaline (NA); elevates brain NA concentrations.

Analysis Note

Supplied as a mixture of erythro and threo isomers


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library