Skip to Content
Merck
All Photos(1)

Key Documents

JVM-13

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

human blood

packaging

tube of 5 μg 96090516-DNA-5UG
pkg of vial of cells 96090516-1VL

growth mode

Suspension

karyotype

46XY,+t(8,?13)(p12-21,?q13)+(8,?3)(p12-21,?q21)+I

morphology

Lymphoblastoid

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian B-prolymphocytic leukaemia, EBV transformed

Cell Line Description

Derived from the peripheral blood of a human Caucasian male patient with prolymphocytic leukaemia. Cells have been subsequently transformed by treatment with EBV and TPA (12-0-tetradecanoylphorbol-13-acetate). JVM-13 cells express surface-membrane and cytoplasmic lambda light chain immunoglobulin. Immunological markers detected were HLA-DR, CD19, CD20, CD21, FMC7, CD38, CD71 and CD9.

Application

Study of B cell leukaemia

DNA Profile

STR-PCR Data: Amelogenin: X,Y
CSF1PO: 10,12
D13S317: 9,11
D16S539: 11
D5S818: 11
D7S820: 9
THO1: 6,7
TPOX: 10,11
vWA: 16,18

Culture Medium

RPMI 1640 + 4mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service