Skip to Content
Merck
All Photos(1)

Key Documents

CL-S1

90110502, mouse breast, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

CL-S1, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., 90110502

biological source

mouse breast

packaging

tube of 5 μg 90110502-DNA-5UG
pkg of vial of cells 90110502-1VL

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

A-type and C-type virus particles, plasminogen activator (tPA)

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse BALB/c mammary alveolar nodules, pre-neoplastic

Cell Line Description

Derived from pre-malignant mammary tissue of a BALB/c mouse. Produce significant amounts of C-type and A-type virus particles and low amounts of plasminogen activator.

Application

Non malignant mammary cell studies.

Culture Medium

DMEM + 2mM Glutamine + 10μg/ml Insulin + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split semi-confluent cultures 1:2 to 1:4 i.e. seeding at 3-5x10,000 cells/cm2 using 0.25% trypsin; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service