Skip to Content
Merck
All Photos(1)

Key Documents

Y13-259

89111412, Reactivity: Anti-p21 Protein, Harvey Murine Sarcoma Virus, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Rat x Rat Hybridoma

packaging

tube of 5 μg 89111412-DNA-5UG
pkg of vial of cells 89111412-1VL

ImmunoDon

Osborne-Mendel rat spleen

Myeloma

Y3Ag1.2.3

Reactivity

Anti-p21 Protein, Harvey Murine Sarcoma Virus

technique(s)

cell culture | mammalian: suitable

isotype

IgG1

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Rat Y3Ag1.2.3 x Rat Osborne-Mendel spleen

Cell Line Description

The antibody reacts to p21 encoded by Ha-MuSV v-ras gene. Cross-reacts to p21 encoded by Kirsten MuSV v-ras gene, p21 encoded by BALB-SV and 29kDa gag-ras fusion protein encoded by Ra-SV. Recognises p21 species in some uninfected rodent, carnivore and primate lines.

Immunogen

Ha-MuSV transformed syngeneic NRK cells

Culture Medium

DMEM + 2mM glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C. Cells will semi-attach. Shake cells off or use trypsin to remove cells from flask

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service