Skip to Content
Merck
All Photos(1)

Key Documents

Ehrlich-Lettre Ascites strain E cell line

87032503, mouse unknown/unspecified, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

Ehrlich-Lettre Ascites strain E cell line, from mouse unknown/unspecified, 87032503

biological source

mouse unknown/unspecified

growth mode

Adherent

karyotype

2n = 40

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Looking for similar products? Visit Product Comparison Guide

Cell Line Origin

Mouse carcinoma

Cell Line Description

Established from the explant of a 7 day old tumour from the parental Ehrlich-Lettre Ascites carcinoma. Prior to development, strain E was subjected to a schedule of seven mouse passages and six in vitro passages over a period of 17 months. The cells exhibit an unusually high mean chromosome number and possess the marker chromosomes present in the parental Ehrlich tumour. This cell line may be of use in comparative testing of anti-tumour agents in vivo and in vitro.

Application

Anti-tumour test and tumourigenicity studies

Culture Medium

NCTC 135 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:3 i.e. seeding at 3-5x10,000 cells/cm2using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C. Media change 3 times a week.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service