Skip to Content
Merck
All Photos(1)

Key Documents

OX-39

86100905, Reactivity: Anti-CD25, rat

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Mouse x Mouse Hybridoma

ImmunoDon

BALB/c mouse spleen

Myeloma

NS0/1

Reactivity

Anti-CD25, rat

technique(s)

cell culture | mammalian: suitable

isotype

IgG1

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse NS0/1 x Mouse BALB/c spleen

Cell Line Description

The antibody is directed against the rat IL-2 receptor found on T cell blasts and thymic dendritic cells. The OX-39 antigen has a molecular weight of 45-50kDa.

Immunogen

T blasts from a mixed lymphocyte reaction between purified CD4+ T cells and irradiated spleens

Culture Medium

RPMI 1640 + 2mM glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Seed at 5x10,000 viable cells/ml. Subculture when cells reach 5x100,000-1x1,000,000 cells/ml (usually after 3-4 days); 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service