Skip to Content
Merck
All Photos(1)

Key Documents

SC-1

86060301, mouse embryo, Fibroblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
biological source:
mouse embryo
growth mode:
Adherent
karyotype:
Not specified
morphology:
Fibroblast
products:
Not specified
receptors:
Not specified

Product Name

SC-1, 86060301

biological source

mouse embryo

growth mode

Adherent

karyotype

Not specified

morphology

Fibroblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse feral embryo

Cell Line Description

Sensitive to both N- and B- trophic viruses. Cells exhibit a greater sensitivity than other mouse lines to MuLV strains and to a limited extent support the replication of xenotropic MuLV. This cell line has been mycoplasma eradicated at ECACC.

Application

Virus studies: N- and B- trophic viruses, MuLV

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:6 i.e. seeding at 2-5x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service