Skip to Content
Merck
All Photos(1)

Key Documents

HCA-7 Colony 11

9071506, human colon, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

HCA-7 Colony 11, 09071506

biological source

human colon

packaging

tube of 5 μg 09071506-DNA-5UG
pkg of vial of cells 09071506-1VL

growth mode

Adherent

karyotype

Near diploid

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human colorectal adenocarcinoma

Cell Line Description

Subpopulations isolated from the HCA-7 cell line.

DNA Profile

STR-PCR Data:

Amelogenin: X
CSF1PO: 11,14
D13S317: 9
D16S539: 10,12
D5S818: 9,12
D7S820: 11,12
THO1: 6,7
TPOX: 8,10
vWA: 14,16

Culture Medium

DMEM (High glucose Sigma D6546) + 2 mM Glutamine + 10% Fetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:5 to 1:10 weekly using 0.05% trypsin/EDTA; 7.5% CO2; 37 °C. Suggested seeding density 2-4x10,000 cells/cm2.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service