Skip to Content
Merck

SP2/0-Ag14 (AC-free)

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 8060101, mouse unknown/unspecified, Lymphoblast

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41106514
Biological source:
mouse unknown/unspecified
Relevant disease(s):
cancer
Growth mode:
Suspension
Karyotype:
Not specified
Morphology:
Lymphoblast
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse unknown/unspecified

packaging

tube of 5 μg 08060101-DNA-5UG, pkg of vial of cells 08060101-1VL

growth mode

Suspension

karyotype

Not specified

morphology

Lymphoblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Application

Fusion partner

Biochem/physiol Actions

Mouse x mouse hybridoma non-secreting, serum-free, animal component (AC) free
Sp2/0-Ag14 cell line (Sigma Catalogue number. 85072401) adapted to grow in animal component free medium (EX-CELL Sp2/0 Serum-Free Medium for Sp2/0 Cells Chemically Defined, Sigma cat no 14660C). Sp2/0-Ag14 is a non-Ig-secreting or synthesising line derived from a cell line created by fusing a BALB/c mouse spleen cell and the mouse myeloma P3X63Ag8. Resistant to 8-azaguanine at 20ug/ml and does not survive in HAT containing media. Can be used as a fusion partner for generating hybridomas.

Preparation Note

EX-CELL Sp2/0 Serum-Free Medium (Sigma cat no. 14660C) + 8mM Glutamine. When freezing cells down use 50:50 fresh culture medium:conditioned medium plus 10% DMSO)
Viability may be poor on resuscitation and may initially decrease further. Full recovery may take up to 2 weeks. A centrifugation step to remove the cryoprotectant is essential. Rapidly thaw the frozen ampoule in a water bath at 37°C for 1-2 minutes. Transfer the contents to a centrifuge tube and slowly add 5-10ml of pre-warmed growth media. Remove a sample for counting. Centrifuge at 100g for 2-3 minutes to pellet cells and seed at a relatively high density of 5-7 x 105 cells/ml. Leave culture flask upright and observe regularly until viable proliferating cells are seen. Once established use a split ratio of 1:2 approximately every 4 to 5 days; 5% CO2; 37°C..

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
This cell line is part of the European Collection of Authenticated Cell Cultures (ECACC), an international repository managed by the United Kingdom Health Security Agency (UKHSA). No licensing agreement is required when either this cell line or the DNA extracted from it are used for internal research purposes only. Any other use of these products is prohibited without the express written permission of UKHSA. Inquiries regarding authorized use of this cell line or its genomic DNA may be directed to culturecollections@ukhsa.gov.uk.




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library