PE/CA-PJ49
00060606, human tongue, squamous cell carcinoma. Epithelial-like morphology.
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
Recommended Products
product name
PE/CA-PJ49, 00060606
biological source
human mouth
growth mode
Adherent
karyotype
Not specified
morphology
Epithelial-like
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Human oral squamous cell carcinoma
Cell Line Description
The squamous carcinoma cell line PE/CA-PJ49 has been obtained from a caucasian male of 57 years old.
Application
The cells can be used in in vitro invasion models for oral squamous epithelial carcinomas to evaluate turnaround stromal cell properties, as well as extra-cellular matrix.
Culture Medium
Subculture Routine
Split sub-confluent cultures (70-80%) 1:4-1:8 ie. seeding at 1-3 x10,000 cells/cm2 using 0.25% Tyrpsin/EDTA, 37°C, 5% CO2
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service