Skip to Content
Merck

C8721

Cetrinix supplement

suitable for microbiology

Synonym(s):

C-N supplement

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.85
UNSPSC Code:
41171614
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


agency

according to ISO 16266:2006

Quality Segment

sterility

sterile

form

powder

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
environmental
food and beverages, microbiology

storage temp.

2-8°C

suitability

Pseudomonas spp.

Application

An antibiotic supplement recommended for the selective isolation of Pseudomonas species. One vial is sufficient for 500 mL medium.

Other Notes

Components per vial in mg:
Cetrimide, 100.00
Nalidixic Acid, 7.50


signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library