Skip to Content
Merck
All Photos(1)

Key Documents

B3686

Sigma-Aldrich

BAY U6751 hydrate

solid, ≥98% (HPLC)

Synonym(s):

4-(2-Chlorophenyl)-1-ethyl-1,4-dihydro-6-methyl-2,3,5-pyridinetricarboxylic acid 5-isopropyl ester disodium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H20ClNNa2O6 · xH2O
CAS Number:
Molecular Weight:
451.81 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

storage condition

protect from light

color

white

solubility

H2O: >8 mg/mL

originator

Bayer

storage temp.

2-8°C

SMILES string

[Na+].[Na+].[H]O[H].CCN1C(C)=C(C(c2ccccc2Cl)C(C([O-])=O)=C1C([O-])=O)C(=O)OC(C)C

InChI

1S/C20H22ClNO6.2Na.H2O/c1-5-22-11(4)14(20(27)28-10(2)3)15(12-8-6-7-9-13(12)21)16(18(23)24)17(22)19(25)26;;;/h6-10,15H,5H2,1-4H3,(H,23,24)(H,25,26);;;1H2/q;2*+1;/p-2

InChI key

QECLOEJXLYHXNF-UHFFFAOYSA-L

Biochem/physiol Actions

BAY W1807, the active metabolite of BAY R3401, inhibits muscle glycogen phosphorylase a and b. In gel-filtered liver extracts, racemic BAY U6751 (containing active BAY W1807) was tested for inhibition of phosphorylase in the glycogenolytic (in which only phosphorylase a is active). In liver extracts, BAY U6751 (0.9-36 μmol/L) inhibited glycogen synthesis by phosphorylase b (notwithstanding the inclusion of AMP), but not by phosphorylase a. Inhibition of phosphorylase-a-catalyzed glycogenolysis was partially relieved by AMP (500 μmol/L). BAY U6751 facilitated phosphorylase-a dephosphorylation. Isolated hepatocytes and perfused livers were tested for BAY R3401-induced changes in phosphorylase-a:b ratios and glycogenolytic output.

Features and Benefits

This compound was developed by Bayer. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Caution

Light sensitive. Store in amber vials.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service