Skip to Content
Merck
All Photos(2)

Key Documents

AV54293

Sigma-Aldrich

Anti-FUCA1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-α-L-Fucosidase 1, tissue

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

54 kDa

species reactivity

rat, mouse, pig, bovine, human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... FUCA1(2517)

Immunogen

Synthetic peptide directed towards the N terminal region of human FUCA1

Application

Anti-FUCA1 antibody produced in rabbit is suitable for western blotting at a concentration of 1μg/mL.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

FUCA1 gene encodes a lysosomal enzyme, α-L-fucosidase mapped on to chromosome 1 at position 1p34.1-1p36. α-L-fucosidase plays a crucial role in hydrolyzing the fucose-containing glycoproteins and glycolipids. Presence of α-L-fucosidase in preoperative serum serves as a prognostic indicator for hepatocellular carcinoma. Mutation in FUCA1 gene leads to fucosidosis, an autosomal recessive lysosomal storage disease.

Sequence

Synthetic peptide located within the following region: PSPVSWNWNSKDVGPHRDLVGELGTALRKRNIRYGLYHSLLEWFHPLYLL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nobuo Tsuchida et al.
International journal of oncology, 50(6), 2043-2048 (2017-04-26)
Fucose residues of cell surface glycans, which play important roles in growth, invasion and metastasis, are added by fucosyltransferases (FUTs) and removed by α-L-fucosidases (FUCAs). By the differential display method, we isolated a 3' non-coding region of α-L-fucosidase-1 (FUCA1) (a gene
K Wang et al.
British journal of cancer, 110(7), 1811-1819 (2014-02-27)
Preoperative alpha-L-fucosidase (AFU) has been used as a diagnostic biomarker for hepatocellular carcinoma (HCC), but its role as a prognostic predictor after partial hepatectomy has not been well defined. The study aimed to investigate the prognostic significance of preoperative serum
Chiaki Nagai-Okatani et al.
PloS one, 11(6), e0150210-e0150210 (2016-06-10)
Targeted proteomics focusing on post-translational modifications, including glycosylation, is a useful strategy for discovering novel biomarkers. To apply this strategy effectively to cardiac hypertrophy and resultant heart failure, we aimed to characterize glycosylation profiles in the left ventricle and plasma
J K Darby et al.
American journal of human genetics, 43(5), 749-755 (1988-11-01)
Human alpha-L-fucosidase is a lysosomal enzyme responsible for hydrolysis of alpha-L-fucoside linkages in fucoglycoconjugates. A single gene, FUCA 1, located on chromosome 1p34.1-1p36.1 encodes for alpha-L-fucosidase activity. To gain insight into the nature of the molecular defects leading to fucosidosis
M Williamson et al.
Journal of medical genetics, 30(3), 218-223 (1993-03-01)
Fucosidosis is a rare, autosomal recessive, lysosomal storage disease, resulting from a deficiency of the enzyme alpha-fucosidase (EC 3.2.1.51). It is characterised clinically by progressive mental and motor deterioration, growth retardation, coarse facies, and often recurrent infections, but the course

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service