AMAB91847
Anti-CD44 antibody produced in mouse

Prestige Antibodies® Powered by Atlas Antibodies, clone CL13324, purified by using Protein A, buffered aqueous glycerol solution
Synonym(s):
CD44R,CSPG8,HCELL,IN,MC56,MDU2,MDU3,MIC4,Pgp1
About This Item
Recommended Products
biological source
mouse
Quality Level
conjugate
unconjugated
antibody product type
primary antibodies
clone
CL13324, monoclonal
product line
Prestige Antibodies® Powered by Atlas Antibodies
form
buffered aqueous glycerol solution
purified by
using Protein A
species reactivity
human
enhanced validation
orthogonal RNAseq
Learn more about Antibody Enhanced Validation
technique(s)
immunohistochemistry: 1:2500-1:5000
western blot: 1 μg/mL
isotype
IgG2b
Ensembl | human accession no.
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... CD44(960)
Immunogen
Application
Features and Benefits
Physical form
Storage and Stability
Legal Information
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service