Skip to Content
Merck
All Photos(5)

Key Documents

AMAB90854

Sigma-Aldrich

Monoclonal Anti-RHOT1 antibody produced in mouse

Prestige Antibodies® Powered by Atlas Antibodies, clone CL1095, purified immunoglobulin, buffered aqueous glycerol solution

Synonym(s):

ARHT1, FLJ11040, MIRO-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

CL1095, monoclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

isotype

IgG1

Ensembl | human accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RHOT1(55288)

General description

Miro1/RHOT1 (ras homolog family member T1) is an outer mitochondrial membrane (OMM) protein. It has a transmembrane domain, two GTPase domains and two Ca2+-sensing EF-hand domains. RHOT1 is located on human chromosome 17q11.

Immunogen

ras homolog family member T1, recombinant protein epitope signature tag (PrEST)

Sequence
THIVDYSEAEQSDEQLHQEISQANVICIVYAVNNKHSIDKVTSRWIPLINERTDKDSRLPLILVGNKSDLVEYSSMETILPIMNQYTEIE

Epitope
Binds to an epitope located within the peptide sequence ERTDKDSRLP as determined by overlapping synthetic peptides.

Application

Monoclonal Anti-RHOT1 antibody has been used in Immunoprecipitation.

Biochem/physiol Actions

Miro1/RHOT1 (ras homolog family member T1) regulates mitochondrial trafficking and distribution. The Rho GTPase Miro-1 plays a crucial role in the modulation of mitochondrial morphogenesis and acts as a calcium-dependent sensor for the control of mitochondrial motility.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71907

Physical form

Phospate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evidence-based genomic diagnosis characterized chromosomal and cryptic imbalances in 30 elderly patients with myelodysplastic syndrome and acute myeloid leukemia
Bajaj R, et al.
Molecular Cytogenetics, 4(1), 3-3 (2011)
K27 ubiquitination of the mitochondrial transport protein Miro is dependent on serine 65 of the Parkin ubiquitin ligase.
Birsa N, et al.
Test, jbc-M114 (2014)
DISC1-dependent regulation of mitochondrial dynamics controls the morphogenesis of complex neuronal dendrites
Norkett R, et al.
The Journal of Biological Chemistry, 291(2), 613-629 (2016)
Miro-1 links mitochondria and microtubule Dynein motors to control lymphocyte migration and polarity
Morlino G, et al.
Molecular and cellular biology, MCB-01177 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service