Skip to Content
Merck
All Photos(1)

Key Documents

61305

Sigma-Aldrich

Penicillinase from Bacillus cereus

lyophilized, powder, white, ~13 U/mg

Synonym(s):

β-Lactamase I from Bacillus cereus, β-Lactamase II, Cephalosporinase, Penicillin amido-β-lactam hydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54
Pricing and availability is not currently available.

biological source

Bacillus sp. (Bacillus cereus)

Quality Level

form

powder

quality

lyophilized

specific activity

~13 U/mg

mol wt

Mr ~30000

technique(s)

activity assay: suitable

color

white

suitability

suitable for component for culture media

application(s)

diagnostic assay manufacturing

foreign activity

β-lactamase II ≤2%

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Penicillinase from Bacillus cereus is both an extracellular and cell-bound enzyme. It has a single polypeptide chain and has a molecular weight of 31 kDa. It lacks cysteine in the protein sequence.[1]

Application

Penicillinase from Bacillus cereus may be used in the preparation of biosensor for amperometric detection of hydrolysis of penicillin.[2]
Used in the production of penicillin.

Biochem/physiol Actions

Penicillinase specifically catalyzes the hydrolysis of β-lactam ring of penicillin.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Unit Definition

One unit will hydrolyze 1.0 μmole of indicated substrate per min at pH 7.0 at 25 °C. Sold as benzylpenicillin units. This International Unit (using benzylpenicillin as substrate) is approximately equal to 600 Levy or 75 Pollock units.

Other Notes

Used to destroy β-lactam antibiotics in body fluids or culture media. For the determination of penicillin[3][4]; In the coupled enzyme assay for isopenicillin N synthetase[5]; ELISA of estradiol[6]; Penicillinase optodes[7]

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J E Baldwin et al.
Analytical biochemistry, 145(1), 183-187 (1985-02-15)
The development of a coupled enzyme assay for the determination of isopenicillin N synthetase activity in purified extracts from Cephalosporium acremonium was described. Isopenicillin N formed from its precursor, delta-(L-alpha-aminoadipyl)-L-cysteinyl-D-valine (ACV), by the synthetase was hydrolyzed by beta-lactamase I to
W. Hobel et al.
Biosensors And Bioelectronics, 7, 549-549 (1992)
T. Scheper et al.
Analytica Chimica Acta, 163, 111-111 (1984)
Flow injection determination of penicillins using immobilized penicillinase in a single bead string reactor.
R Gnanasekaran et al.
Analytical chemistry, 57(6), 1005-1009 (1985-05-01)
Properties of penicillinase from Bacillus cereus 569
Imsande J, et al.
The Journal of Biological Chemistry, 245(9), 2205-2212 (1970)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service