27674
Collagenase-Substrate Test Substance
powder
Synonym(s):
Gly-Pro-Ala
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C10H17N3O4
CAS Number:
Molecular Weight:
243.26
Beilstein:
5288874
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32
Recommended Products
Product Name
Collagenase-Substrate Test Substance (for quantitative Collagenase-Determination),
form
powder
Quality Level
shipped in
wet ice
storage temp.
−20°C
SMILES string
C[C@H](NC(=O)[C@@H]1CCCN1C(=O)CN)C(O)=O
InChI
1S/C10H17N3O4/c1-6(10(16)17)12-9(15)7-3-2-4-13(7)8(14)5-11/h6-7H,2-5,11H2,1H3,(H,12,15)(H,16,17)/t6-,7-/m0/s1
InChI key
GGLIDLCEPDHEJO-BQBZGAKWSA-N
Related Categories
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Tadashi Hatanaka et al.
Journal of enzyme inhibition and medicinal chemistry, 29(6), 823-828 (2014-03-22)
The collagen tripeptide fragments Gly-Ala-Hyp, Gly-Pro-Ala and Gly-Pro-Hyp were generated by hydrolyzing collagen from pig-skin, cattle-skin, fish-scales and chicken-feet, respectively, with Streptomyces collagenase. Collagenase treatment increased the concentration of tripeptides in the hydrolysates by 13-15% (w/w). Of the three peptides
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service