19160
SOD Assay Kit
sufficient for 500 tests
Synonym(s):
Superoxide dismutase Assay Kit
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
usage
sufficient for 500 tests
Quality Level
availability
not available in USA
technique(s)
UV/Vis spectroscopy: suitable
suitability
corresponds for UV test (WST reagent, WST formazan)
use tested
storage temp.
2-8°C
General description
The kit contains all reagents and solutions required for determining superoxide dismutase activity in an indirect assay method based on xanthine oxidase and a novel color reagent. The chemical and biochemical properties of the color reagent used in this kit guarantee a very convenient application and linearity of test results over a broad range.
Application
SOD Assay Kit has been used for the determination of superoxide dismutase activity in mammalian cells.
Biochem/physiol Actions
Superoxide dismutases are enzymes responsible for the elimination of superoxide radical (O2-), thereby protecting cells from free radicals-associated damages. They catalyze the dismutation of two superoxide radicals to hydrogen peroxide and oxygen.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Resp. Sens. 1
Storage Class Code
10 - Combustible liquids
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Superoxide dismutase as a target for the selective killing of cancer cells.
Nature, 407, 390-390 (2000)
Extracellular superoxide dismutase in biology and medicine.
Free Radical Biology & Medicine, 35, 236-236 (2003)
Atg7 Knockdown Augments Concanavalin A-Induced Acute Hepatitis through an ROS-Mediated p38/MAPK Pathway.
PLoS ONE, 11, e0149754-e0149754 (2016)
Frontiers in plant science, 8, 830-830 (2017-07-01)
The negative effects of the ongoing climate change include unusual prolonged droughts and increased salinity pressures on the agricultural lands. Consequently, crops are facing unprecedented environmental pressure, and this calls for more research toward controlling such major stresses. The current
Chronic ethanol consumption induces erectile dysfunction: Role of oxidative stress.
Life Sciences, 141, 44-44 (2015)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service