11012005
OACP4 C
human gastric cardia, pleiomorphic: epithelial and fibroblastoid cells, cytoplasmic protrusions
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
product name
OACP4 C,
biological source
human gastric cardia
Quality Level
description
Gastric cardia adenocarcinoma
growth mode
adherent
karyotype
Aneuploid 53-57 chromosomes
morphology
Pleiomorphic: both epithelial and fibroblastoid cells with cytoplasmic protrusions
technique(s)
cell culture | mammalian: suitable
shipped in
dry ice
storage temp.
−196°C
Cell Line Description
OACP4 C was established from a primary tumour located in gastric cardia of a 55 year-old Caucasian male. The patient showed lymph node involvement with pleural metastases. OACP4 C was found to be tumourigenic in NMRI (female nude mice or NOD-SCID mice. The Y chromosome could not be detected in this cell line by short tandem repeat (STR)-PCR analysis when tested at ECACC. It is a known phenomenon that due to the increased genetic instability of cancer cell lines the Y chromosome can be rearranged or lost resulting in lack of detection. The cell line is identical to the source provided by the depositor based on the STR-PCR analysis.
Cultures derived from ECACC stocks of this cell line have been whole genome sequenced (Contino, et al 2016) confirming the presence of many of the known mutations that drive oesophageal cancer.
Cultures derived from ECACC stocks of this cell line have been whole genome sequenced (Contino, et al 2016) confirming the presence of many of the known mutations that drive oesophageal cancer.
DNA Profile
STR-PCR Data:
Amelogenin: X
CSF1PO: 11
D13S317: 12
D16S539: 12
D5S818: 9
D7S820: 9,11
THO1: 9
TPOX: 11
vWA: 16
Amelogenin: X
CSF1PO: 11
D13S317: 12
D16S539: 12
D5S818: 9
D7S820: 9,11
THO1: 9
TPOX: 11
vWA: 16
Culture Medium
Subculture Routine
Split sub-confluent cultures (70-80%) up to 1:5 i.e. seeding at 1-3 x 10,000 cells/cm² using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Cultures from PHE Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.
Citation
If use of this culture results in a scientific publication, it should be cited in the publication as: OACP4 C (ECACC 11012005)
Disclaimer
RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service