Skip to Content
Merck
All Photos(1)

Key Documents

A1010

Sigma-Aldrich

Arsenic(III) oxide

ReagentPlus®, ≥99.0%

Synonym(s):

Arsenic trioxide, Arsenous acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
As2O3
CAS Number:
Molecular Weight:
197.84
EC Number:
MDL number:
UNSPSC Code:
12171602
PubChem Substance ID:
NACRES:
NA.55
Assay:
≥99.0%
form:
powder

vapor pressure

0 hPa ( 66 °C)

Quality Level

product line

ReagentPlus®

Assay

≥99.0%

form

powder

storage temp.

room temp

SMILES string

O=[As]O[As]=O

InChI

1S/As2O3/c3-1-5-2-4

InChI key

IKWTVSLWAPBBKU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Arsenic(III) oxide, also known as arsenic trioxide, is an amphoteric oxide that serves as a raw material for the synthesis of several arsenic-based compounds. It is utilized to decolorize glasses and enamels. Its efficacy in treating in acute promyelocytic leukaemia (APL) has been investigated.

Application

Arsenic(III) oxide was used in a study detailing the effect of arsenic trioxide in combination with γ-irradiation to elucidate the hemoglobin (Hb).

Biochem/physiol Actions

Very potent agent against acute promyelocytic leukemia (APL). Causes skin, lung and bladder cancer.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A - Eye Dam. 1 - Skin Corr. 1B - STOT RE 1

Target Organs

Respiratory system,Cardio-vascular system,Gastrointestinal tract

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Lengfelder et al.
Leukemia, 26(3), 433-442 (2011-09-10)
Arsenic trioxide (ATO) is presently the most active single agent in the treatment of acute promyelocytic leukemia (APL). This review provides insights into the mode of action and the pharmacological properties of ATO, and summarizes the most relevant results of
Combined effect of arsenic trioxide and radiation on physical properties of hemoglobin biopolymer.
Saad-El-Din AA, et al.
Journal of Radiation Research and Applied Sciences, 7(2), 411-416 (2014)
Wilson H Miller et al.
Cancer research, 62(14), 3893-3903 (2002-07-19)
Arsenic trioxide has shown substantial efficacy in treating both newly diagnosed and relapsed patients with acute promyelocytic leukemia (APL). As a single agent, it induces complete remissions, causing few adverse effects and only minimal myelosuppression. These successes have prompted investigations
Dikshith TSS
Handbook of Chemicals and Safety, 92-92 (1992)
Z X Shen et al.
Blood, 89(9), 3354-3360 (1997-05-01)
The therapeutic effect of arsenic trioxide (As2O3) in the treatment of acute promyelocytic leukemia (APL) was evaluated among 15 APL patients at relapse after all-trans retinoic acid (ATRA) induced and chemotherapy maintained complete remission (CR). As2O3 was administered intravenously at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service