Skip to Content
Merck
All Photos(1)

Key Documents

72285

Sigma-Aldrich

Nickel(II) sulfate heptahydrate

purum p.a., crystallized, ≥99.0% (KT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NiSO4 · 7H2O
CAS Number:
Molecular Weight:
280.86
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99.0% (KT)
form:
crystals

grade

purum p.a.

Quality Level

Assay

≥99.0% (KT)

form

crystals

quality

crystallized

density

1.948 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤50 mg/kg

cation traces

Ca: ≤50 mg/kg
Cd: ≤50 mg/kg
Co: ≤500 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤100 mg/kg
Na: ≤100 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

[Ni++].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[O-]S([O-])(=O)=O

InChI

1S/Ni.H2O4S.7H2O/c;1-5(2,3)4;;;;;;;/h;(H2,1,2,3,4);7*1H2/q+2;;;;;;;;/p-2

InChI key

OGKAGKFVPCOHQW-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Nickel(II) sulfate hexahydrate is a hydrated nickel salt. Its thermal decomposition in the presence of air and helium has been studied. Its crystals exist in monoclinic and tetragonal polymorphic forms.

Application

Nickel(II) sulfate hexahydrate may be used in the preparation of NiFe2O4 nanoparticles (NPs), required for the fabrication of a glucose biosensor.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A Inhalation - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Inhalation

Target Organs

respiratory tract irritation

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amperometric glucose biosensor based on NiFe2O4 nanoparticles and chitosan.
Luo L, et al.
Sensors and Actuators B, Chemical, 145(1), 293-298 (2010)
Mechanism and kinetics of thermal decomposition of nickel (II) sulfate (VI) hexahydrate.
Tomaszewicz E and Kotfica M.
Journal of Thermal Analysis and Calorimetry, 77(1), 25-31 (2004)
Polymorphism of nickel sulfate hexahydrate.
Angel RJ and Finger LW.
Acta Crystallographica Section C, Crystal Structure Communications, 44(11), 1869-1873 (1988)
Xiaohua Xu et al.
Particle and fibre toxicology, 9, 40-40 (2012-11-07)
It has been well recognized that toxicity of fine ambient air particulate matter (PM(2.5)) may depend on its chemical constituents, including components such as soluble metals that may theoretically exert distinctive effects. We have recently demonstrated an important effect of
A Tammaro et al.
International journal of immunopathology and pharmacology, 25(2), 519-521 (2012-06-16)
Nickel deficiency leads to reduced iron content in organs and to reduced hemoglobin levels and haematocrit counts. We carried out a clinical trial of oral hyposensitization therapy with low doses of nickel on a group of 20 patients affected by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service