Skip to Content
Merck
All Photos(1)

Key Documents

270652

Supelco

Carbon tetrachloride

suitable for HPLC, ≥99.9%

Synonym(s):

Tetrachloromethane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CCl4
CAS Number:
Molecular Weight:
153.82
Beilstein:
1098295
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99.9%
technique(s):
HPLC: suitable
application(s):
food and beverages
bp:
76-77 °C (lit.)
vapor pressure:
143 mmHg ( 30 °C)
91 mmHg ( 20 °C)
Pricing and availability is not currently available.

vapor density

5.32 (vs air)

Quality Level

vapor pressure

143 mmHg ( 30 °C)
91 mmHg ( 20 °C)

Assay

≥99.9%

form

liquid

purified by

glass distillation

availability

not available in EU

technique(s)

HPLC: suitable

impurities

≤0.01% water

evapn. residue

<0.0004%

refractive index

n20/D 1.460 (lit.)

bp

76-77 °C (lit.)

mp

−23 °C (lit.)

density

1.594 g/mL at 25 °C (lit.)

λ

H2O reference

UV absorption

λ: 265 nm Amax: 1.0
λ: 270 nm Amax: 0.30
λ: 280 nm Amax: 0.07
λ: 290 nm Amax: 0.02
λ: 300-400 nm Amax: 0.01

application(s)

food and beverages

SMILES string

ClC(Cl)(Cl)Cl

InChI

1S/CCl4/c2-1(3,4)5

InChI key

VZGDMQKNWNREIO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This solvent is glass distilled, submicron filtered and subjected to rigorous specification testing to provide lot-to-lot consistency.
Please view www.aldrich.com/epaods regarding the EPA?s request for application information of Ozone Depleting Substances.

Application

Designed for use with HPLC instrumentation and organic synthesis applications.
Please view www.aldrich.com/epaods regarding the EPA′s request for application information of Ozone Depleting Substances

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Carc. 2 - Ozone 1 - Skin Sens. 1B - STOT RE 1 Inhalation

Target Organs

Liver,Kidney

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Keywan Mortezaee et al.
Canadian journal of physiology and pharmacology, 1-12 (2015-11-18)
Melatonin is known for being beneficial in targeting liver diseases. This study aimed to investigate whether melatonin post-treatment is capable of rat carbon tetrachloride (CCl
Jae Hyuk Lee et al.
Journal of the American Chemical Society, 135(8), 3255-3261 (2013-02-05)
Molecules are often born with high energy and large-amplitude vibrations. In solution, a newly formed molecule cools down by transferring energy to the surrounding solvent molecules. The progression of the molecular and solute-solvent cage structure during this fundamental process has
Marc Mejias et al.
Gut, 64(4), 657-666 (2014-05-23)
Antiangiogenic strategies have been proposed as a promising new approach for the therapy of portal hypertension and chronic liver disease. Pigment epithelium-derived factor (PEDF) is a powerful endogenous angiogenesis inhibitor whose role in portal hypertension remains unknown. Therefore, we aimed
Samar Basu
Toxicology, 189(1-2), 113-127 (2003-06-25)
Hepatic injury through carbon tetrachloride (CCl(4)) induced lipid peroxidation is well known and has been extensively used in the experimental models to understand the cellular mechanisms behind oxidative damage and further to evaluate the therapeutic potential of drugs and dietary
Solvent toxicology: recent advances in the toxicology of benzene, the glycol ethers, and carbon tetrachloride.
G F Kalf et al.
Annual review of pharmacology and toxicology, 27, 399-427 (1987-01-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service