Skip to Content
Merck
All Photos(4)

Key Documents

205222

Sigma-Aldrich

Sodium cyanide

ACS reagent, ≥95.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaCN
CAS Number:
Molecular Weight:
49.01
Beilstein:
3587243
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NB.24
Assay:
≥95.0%
grade:
ACS reagent
form:
crystalline powder
powder, crystals or chunks
solubility:
water: soluble(lit.)

grade

ACS reagent

Quality Level

vapor density

1.7 (vs air)

vapor pressure

1 mmHg ( 817 °C)

Assay

≥95.0%

form

crystalline powder
powder, crystals or chunks

pH

11-12 (25 °C, 49 g/L)

mp

563.7 °C (lit.)

solubility

water: soluble(lit.)

anion traces

S2-: ≤0.005%
SCN-: ≤0.02%
chloride (Cl-): ≤0.15%
phosphate (PO43-): ≤0.02%
sulfate (SO42-): ≤0.05%

cation traces

Fe: ≤0.005%
Pb: ≤0.005%

SMILES string

[Na]C#N

InChI

1S/CN.Na/c1-2;

InChI key

RTVFYQXEHKQMKO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sodium cyanide is colorless sodium salt and its crystals belong to the cubic crystal system. Its aqueous solutions are strongly basic due to the presence of cyanide ion. It can be synthesized by reacting sodium hydroxide with hydrogen cyanide. Sodium cyanide is a key precursor for the synthesis of various organic intermediates. It participates in the RuCl3-mediated oxidative cyanation of tertiary amines.

Application

Sodium cyanide may be employed for the synthesis of the following compounds:
  • α-amino acids
  • Prussian blue (cyanide pigment)
  • nitriles, via reaction with primary and secondary chlorides

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Inhalation - Acute Tox. 1 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Met. Corr. 1 - STOT RE 1

Target Organs

Thyroid

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eagleson M.
Concise Encyclopedia Chemistry, 1037-1037 (1994)
Daniela Gontero et al.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology, 19(9), 1168-1188 (2020-07-18)
Escherichia coli bacteria were determined by in flow cytometry with laser excitation and fluorescence detection applying ultraluminescent core-shell nanoparticles based on Metal Enhanced Fluorescence (MEF). Core-shell nanoparticles consisted of a 40 nm core modified with a silica spacer grafted with
Preparation of Nitriles from Halides and Sodium Cyanide. An Advantageous Nucleophilic Displacement in Dimethyl Sulfoxide1a.
Friedman L and Shechter H.
The Journal of Organic Chemistry, 25(6), 877-879 (1960)
Shun-Ichi Murahashi et al.
Journal of the American Chemical Society, 125(50), 15312-15313 (2003-12-11)
RuCl3-catalyzed oxidative cyanation of tertiary amines with sodium cyanide under molecular oxygen (1 atm) at 60 degrees C gives the corresponding alpha-aminonitriles, which are versatile synthetic intermediates of various compounds such as amino acids and unsymmetrical 1,2-diamines, in excellent yields.
Arne Wick et al.
Water research, 45(12), 3638-3652 (2011-05-17)
The sludge-water distribution of a total of 41 organic micropollutants (9 phenylurea herbicides, 11 triazines, 16 biocides and 5 UV-filters) was investigated in laboratory batch experiments with fresh secondary sludge taken from a municipal WWTP. Sorption kinetics as well as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service