Skip to Content
Merck
All Photos(1)

Key Documents

12634

Sigma-Aldrich

Potassium chlorate

puriss., 99-101%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KClO3
CAS Number:
Molecular Weight:
122.55
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
form:
powder or crystals

grade

puriss.

Quality Level

Assay

99-101%

form

powder or crystals

impurities

≤0.001% heavy metals (as Pb)

pH

5-6.5 (25 °C, 61.3 g/L)

mp

356 °C (lit.)

anion traces

chloride (Cl-): ≤100 mg/kg
sulfate (SO42-): ≤100 mg/kg

cation traces

As: ≤2 mg/kg
Ca: ≤50 mg/kg
Fe: ≤30 mg/kg

SMILES string

[K+].[O-]Cl(=O)=O

InChI

1S/ClHO3.K/c2-1(3)4;/h(H,2,3,4);/q;+1/p-1

InChI key

VKJKEPKFPUWCAS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Crystals of potassium chlorate belong to the monoclinic crystal system. On heating above 400°C, it degrades to give potassium chloride and potassium perchlorate. It has strong oxidizing properties. On exposure to X-rays, oxygen-17 enriched KClO3 generates O3- radical, which was investigated by ESR (Electron Spin Resonance). Irradiation of potassium chlorate with X-ray causes the degradation to generate chloride, chlorite, hypochlorite and occluded oxygen. Significant quantities of dichlorine hexoxide and a trace of chlorine dioxide were also produced.

Application

Potassium chlorate may be used in the preparation of graphite oxide (GO).

Pictograms

Flame over circleSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Ox. Sol. 1

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Single sheet functionalized graphene by oxidation and thermal expansion of graphite.
McAllister MJ, et al.
Chemistry of Materials, 19(18), 4396-4404 (2007)
The Decomposition of Solid Potassium Chlorate by X Rays.
Heal HG.
Canadian Journal of Chemistry, 37(5), 979-987 (1959)
ESR Spectrum of O-3 Trapped in a Single Crystal of Potassium Chlorate.
Schlick S.
J. Chem. Phys. , 56, 654-661 (1972)
Hakan Mutlu et al.
AJNR. American journal of neuroradiology, 24(7), 1396-1398 (2003-08-15)
We present the case of a patient who attempted suicide by ingesting matchstick heads (55% potassium chlorate). The patient presented to the emergency room with loss of consciousness, and MR imaging revealed symmetric hyperintense signal within the deep gray matter
F Khan et al.
Biochimica et biophysica acta, 1481(2), 229-236 (2000-10-06)
Characterization of conformational transition and folding intermediates is central to the study of protein folding. We studied the effect of various alcohols (trifluoroethanol (TFE), butanol, propanol, ethanol and methanol) and salts (K(3)FeCN(6), Na(2)SO(4), KClO(4) and KCl) on the acid-induced state

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service