Skip to Content
Merck
All Photos(1)

Documents

Y0001406

Glucosamine hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

D-(+)-Glucosamine hydrochloride, 2-Amino-2-deoxy-D-glucose hydrochloride, Chitosamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13NO5 · HCl
CAS Number:
Molecular Weight:
215.63
Beilstein:
4157370
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

Agency

EP Reference Standard

API family

glucosamine

manufacturer/tradename

EDQM

mp

190-194 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl.N[C@H]1C(O)O[C@H](CO)[C@@H](O)[C@@H]1O

InChI

1S/C6H13NO5.ClH/c7-3-5(10)4(9)2(1-8)12-6(3)11;/h2-6,8-11H,1,7H2;1H/t2-,3-,4-,5-,6?;/m1./s1

InChI key

QKPLRMLTKYXDST-NSEZLWDYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Glucosamine hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Glucosamine, an amino sugar, is the precursor of the hexosamine biosynthetic pathway leading to the formation of UDP-N-acetylglucosamine (UDP-GlcNAc), which is then used for making glycosaminoglycans, proteoglycans, and glycolipids. D-(+)-Glucosamine inhibits the coaggregation of Candida yeast species with the bacterial strain S. salivarius.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oshra Betzer et al.
ACS nano, 8(9), 9274-9285 (2014-08-19)
A critical problem in the development and implementation of stem cell-based therapy is the lack of reliable, noninvasive means to image and trace the cells post-transplantation and evaluate their biodistribution, final fate, and functionality. In this study, we developed a
Viktor Chesnokov et al.
Cancer cell international, 14, 45-45 (2014-06-17)
We have reported that the glucosamine suppressed the proliferation of the human prostate carcinoma cell line DU145 through inhibition of STAT3 signaling. DU145 cells autonomously express IL-6 and the IL-6/STAT3 signaling is activated. IL-6 receptor subunits are subject to N-glycosylation
Ji-Sun Hwang et al.
Metabolism: clinical and experimental, 64(3), 368-379 (2014-12-18)
This study investigated the potential of glucosamine (GlcN) to affect body weight gain and insulin sensitivity in mice normal and at risk for developing diabetes. Male C57BL/6J mice were fed either chow diet (CD) or a high fat diet (HFD)
Khadijeh Jamialahmadi et al.
Environmental toxicology and pharmacology, 38(1), 212-219 (2014-06-25)
Glucosamine (GlcN) is an important precursor in the biochemical synthesis of glycosylated proteins and lipids in human body. It gains importance because of its contribution to human health and its multiple biological and therapeutic effects. In this study, the in
Hiba Al-Hamidi et al.
Pharmaceutical development and technology, 20(1), 74-83 (2014-01-08)
The solid dispersion technique is one of the most effective methods for improving the dissolution rate of poorly water-soluble drugs; however this is reliant on a suitable carrier and solvent being selected. The work presented explores amino sugars (d-glucosamine HCl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service