Skip to Content
Merck
All Photos(1)

Key Documents

SMB01353

Sigma-Aldrich

7-Aminoclonazepam

≥95% (HPLC)

Synonym(s):

7-Amino-5-(2-chlorophenyl)-1,3-dihydro-2H-1,4-benzodiazepin-2-one, Aminoclonazepam, Chloraminazepam

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12ClN3O
CAS Number:
Molecular Weight:
285.73
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.22

Quality Level

Assay

≥95% (HPLC)

form

powder

InChI

1S/C15H12ClN3O/c16-12-4-2-1-3-10(12)15-11-7-9(17)5-6-13(11)19-14(20)8-18-15/h1-7H,8,17H2,(H,19,20)

InChI key

HEFRPWRJTGLSSV-UHFFFAOYSA-N

General description

7-Aminoclonazepam is a major metabolite of clonazepam. 7-Aminoclonazepam may be used for detection of clonazepam use.

Application

Metabolomics research

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stacy E F Melanson et al.
Clinica chimica acta; international journal of clinical chemistry, 455, 128-133 (2016-01-31)
Clonazepam (CLON) is not only frequently prescribed in addiction management but is also commonly abused. Therefore many addiction clinics perform oral fluid (OF) testing, which unlike urine is not subject to adulteration, to monitor CLON compliance. However, CLON and other
Claudia Vignali et al.
Journal of analytical toxicology, 45(4), e1-e5 (2020-07-17)
Fluvoxamine is a selective serotonin reuptake inhibitor, with a half-life of about 30 hours, that is commonly prescribed in the treatment of depression and obsessive and compulsive disorders. Though its more favorable adverse effect profile in comparison to tricyclic antidepressants, overdosages
Amadeo Pesce et al.
Annals of clinical and laboratory science, 50(3), 321-326 (2020-06-26)
We examined the results of 1.3 million drug tests performed on patients being monitored for compliance with pain medications and substance abuse rehabilitation to determine if the 2016 CDC prescribing guidelines had any impact on opiate benzodiazepine use. We observed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service