Skip to Content
Merck
All Photos(1)

Key Documents

SMB01317

Sigma-Aldrich

Sinapine thiocyanate

Synonym(s):

Sinapine cyanide sulfonate, Sinapine thiocyanate anhydrous

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H24N2O5S
CAS Number:
Molecular Weight:
368.45
UNSPSC Code:
12352205
NACRES:
NA.77

biological source

synthetic

Quality Level

grade

research grade

Assay

≥98%

form

powder

storage condition

(keep sealed and from light)

technique(s)

HPLC: suitable

color

light yellow

storage temp.

−20°C

InChI

1S/C16H23NO5.CHNS/c1-17(2,3)8-9-22-16(19)7-6-12-10-15(21-5)13(18)11-14(12)20-4;2-1-3/h6-7,10-11H,8-9H2,1-5H3;3H

InChI key

NPOZDPAVPFLLCN-UHFFFAOYSA-N

General description

Sinapine thiocyanate is a salt of sinapine, the choline ester of sinapic acid. Sinapine is known to be present in some foods such as cauliflower, and it has been studied as a dietary precursor of TMAO (trimethylamine N-oxide) in rapeseed-derived feed for pigs.

Application

Metabolomics research, microbiome research

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Doria Boulghobra et al.
Redox biology, 34, 101554-101554 (2020-05-29)
When confronted to stress or pathological conditions, the mitochondria overproduce reactive species that participate in the cellular dysfunction. These organelles are however difficult to target with antioxidants. A feature of mitochondria that can be used for this is the negatively
Louis M M Mouterde et al.
Journal of agricultural and food chemistry, 68(26), 6998-7004 (2020-06-03)
Naturally occurring sinapine was successfully synthesized through a proline-mediated Knoevenagel-Doebner condensation in ethanol. This synthetic process involving biobased syringaldehyde, Meldrum's acid, and choline chloride offers a sustainable alternative to the existing low-yield pathways. This two-step strategy gives access to sinapine
Monika Anna Olszewska et al.
Food & function, 11(8), 7231-7244 (2020-08-08)
Broccoli sprouts represent a health-promoting food, rich in antioxidant and anti-inflammatory phytochemicals, among which sulfur compounds are most extensively investigated. In this study, the phenolics of broccoli sprouts (Brassica oleracea var. italica'Cezar') were examined for variability during germination and influence

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service