Skip to Content
Merck
All Photos(1)

Key Documents

SMB00943

Sigma-Aldrich

Monoisobutyl phthalate

≥95%

Synonym(s):

2-(Isobutoxycarbonyl)benzoic acid, MIBP, Mono(2-isobutyl) phthalate, Mono-2-isobutylphthalate, Monoisobutyl phthalic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14O4
CAS Number:
Molecular Weight:
222.24
MDL number:
UNSPSC Code:
12352205
NACRES:
NA.77

biological source

synthetic

Quality Level

Assay

≥95%

form

solid

mol wt

222.24 g/mol

storage condition

(Keep container tightly closed in a dry and well -ventilated place.)

technique(s)

HPLC: suitable

color

off-white to white

storage temp.

2-8°C

SMILES string

O(CC(C)C)C(=O)c1c(cccc1)C(=O)O

InChI

1S/C12H14O4/c1-8(2)7-16-12(15)10-6-4-3-5-9(10)11(13)14/h3-6,8H,7H2,1-2H3,(H,13,14)

InChI key

RZJSUWQGFCHNFS-UHFFFAOYSA-N

General description

Monoisobutyl phthalate, also known as monoisobutyl phtalic acid or MIBP is a synthetic industrial compound capable of disrupting the endocrine system. It can change levels of testosterone, insulin-like growth factor-3 (IGF-3) and follicule stimulating hormone (FSH).

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kayoko Kato et al.
Analytical chemistry, 78(18), 6651-6655 (2006-09-15)
We developed an analytical method using off-line solid-phase extraction (SPE) coupled with on-line SPE and isotope-dilution high-performance liquid chromatography-tandem mass spectrometry (HPLC-MS/MS) to determine the concentrations of phthalate metabolites in human meconium and in semen. First, we used off-line SPE
Manori J Silva et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 805(1), 161-167 (2004-04-29)
We improved our previous analytical method to measure phthalate metabolites in urine as biomarkers for phthalate exposure by automating the solid-phase extraction (SPE) procedure and expanding the analytical capability to quantify four additional metabolites: phthalic acid, mono-3-carboxypropyl phthalate, mono-isobutyl phthalate
Manori J Silva et al.
Archives of toxicology, 79(11), 647-652 (2005-07-05)
In assessment of exposure to environmental contaminants, the use of unconventional matrices is becoming an increasingly important area of research. Saliva is one of the most promising alternative matrices because its collection is easy, noninvasive, and inexpensive. In this study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service