Skip to Content
Merck
All Photos(1)

Key Documents

S2158000

Sulfathiazole

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2-Sulfanilamidothiazole, 4-Amino-N-(2-thiazolyl)benzenesulfonamide, N1-(2-Thiazolyl)sulfanilamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9N3O2S2
CAS Number:
Molecular Weight:
255.32
Beilstein:
226178
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

sulfathiazole

manufacturer/tradename

EDQM

mp

200-202 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Nc1ccc(cc1)S(=O)(=O)Nc2nccs2

InChI

1S/C9H9N3O2S2/c10-7-1-3-8(4-2-7)16(13,14)12-9-11-5-6-15-9/h1-6H,10H2,(H,11,12)

InChI key

JNMRHUJNCSQMMB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Sulfathiazole EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Indra Mani Sharma et al.
IUBMB life, 66(12), 823-834 (2014-12-30)
Bacteria have evolved to survive the ever-changing environment using intriguing mechanisms of quorum sensing (QS). Very often, QS facilitates formation of biofilm to help bacteria to persist longer and the formation of such biofilms is regulated by c-di-GMP. It is
Mohd R Abu Bakar et al.
International journal of pharmaceutics, 414(1-2), 86-103 (2011-05-21)
Since the discovery of sulfathiazole as an antimicrobial agent in 1939, numerous works in the screening for its different polymorphic forms, which is an essential part of drug development, have been conducted and published. These works consequently result in the
Shaojun Yang et al.
The Journal of pharmacology and experimental therapeutics, 352(3), 529-540 (2015-01-01)
Sepiapterin reductase (SPR) catalyzes the reduction of sepiapterin to dihydrobiopterin (BH2), the precursor for tetrahydrobiopterin (BH4), a cofactor critical for nitric oxide biosynthesis and alkylglycerol and aromatic amino acid metabolism. SPR also mediates chemical redox cycling, catalyzing one-electron reduction of
Qing Wang et al.
Analytica chimica acta, 854, 191-201 (2014-12-07)
A hydrophilic stationary phase (SP) was prepared through grafting N-methylene phosphonic acid chitosan on magnesia-zirconia particles (P-CTS-MgO-ZrO2) via Lewis acid-base interaction. The resulting material was characterized by thermogravimetric analysis, Fourier transform infrared spectroscopy, scanning electron microscope and nitrogen adsorption analysis.
Maki Kanda et al.
Journal of AOAC International, 98(1), 230-247 (2015-04-11)
Residues of 37 polar veterinary drugs belonging to six families (quinolones, tetracyclines, macrolides, lincosamides, sulfonamides, and others) in livestock and fishery products were determined using a validated LC-MS/MS method. There were two key points in sample preparation. First, extraction was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service