Skip to Content
Merck
All Photos(1)

Key Documents

PHR1613

Supelco

Levothyroxine

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

L-Thyroxine, 3,3′,5,5″-Tetraiodo-L-thyronine, 3-[4-(4-Hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]-L-alanine, T4

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H2(I)2OC6H2(I)2CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
776.87
Beilstein:
2228515
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. L0570000
traceable to USP 1365000

API family

levothyroxine

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

223 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

N[C@@H](Cc1cc(I)c(Oc2cc(I)c(O)c(I)c2)c(I)c1)C(O)=O

InChI

1S/C15H11I4NO4/c16-8-4-7(5-9(17)13(8)21)24-14-10(18)1-6(2-11(14)19)3-12(20)15(22)23/h1-2,4-5,12,21H,3,20H2,(H,22,23)/t12-/m0/s1

InChI key

XUIIKFGFIJCVMT-LBPRGKRZSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Levothyroxine is a thyroid hormone that is responsible for the physiological actions such as growth, cardiac functionality, and regulation of carbohydrates, lipid, and protein metabolisms.
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Levothyroxine may be used as a pharmaceutical reference standard for the quantification of the analyte in pharmaceutical formulations and bulk dosage forms and oral liquid formulations by chromatography.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

L-Thyroxine (T4) and triiodo-L-thyronine (T3) are iodine-containing hormones produced from thyroglobulin in the thyroid follicular cells. The stimulation of metabolic rate and regulation of growth and development by these hormones appear to be due to their effects on DNA transcription and, thus, protein synthesis.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC1514 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1

Target Organs

Thyroid,Cardio-vascular system,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oral liquid formulation of levothyroxine is stable in breakfast beverages and may improve thyroid patient compliance
Bernareggi A, et al.
Pharmaceutics, 5(4), 621-633 (2013)
Determination of liothyronine and levothyroxine in thyroid preparations by liquid chromatography
Richheimer SL and Jensen CB
Journal of Pharmaceutical Sciences, 75(2), 215-217 (1986)
The separation and determination of liothyronine and levothyroxine in tablets by reversed-phase high performance liquid chromatography
Smith DJ, et al.
Journal of Chromatographic Science, 19(2), 72-78 (1981)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service