NIST2206
Controlled pore glass - BET specific surface area
NIST® SRM® 2206, nominal pore diameter 300 nm
About This Item
Recommended Products
grade
certified reference material
Quality Level
form
granular
packaging
pkg of 5 g
manufacturer/tradename
NIST®
application(s)
glass & ceramic
SMILES string
[Ca+2].[Na+].[P](=O)([O-])([O-])[O-]
InChI
1S/Ca.Na.H3O4P/c;;1-5(2,3)4/h;;(H3,1,2,3,4)/q+2;+1;/p-3
InChI key
QXJJQWWVWRCVQT-UHFFFAOYSA-K
Looking for similar products? Visit Product Comparison Guide
General description
SRM 2206_cert
SRM 2206_SDS
Other Notes
Legal Information
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service