Skip to Content
Merck
All Photos(3)

Key Documents

G6403

Sigma-Aldrich

Glutaraldehyde solution

50% in H2O, suitable for photographic applications

Synonym(s):

Glutaric dialdehyde solution, Pentane-1,5-dial

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
OHC(CH2)3CHO
CAS Number:
Molecular Weight:
100.12
Beilstein:
605390
MDL number:
UNSPSC Code:
12352114
PubChem Substance ID:
NACRES:
NA.21

form

liquid

Quality Level

concentration

50% in H2O

color

colorless

mp

-21  °C ((-6 °F))

solubility

water: soluble

suitability

suitable for photographic applications

SMILES string

[H]C(CCCC([H])=O)=O

InChI

1S/C5H8O2/c6-4-2-1-3-5-7/h4-5H,1-3H2

InChI key

SXRSQZLOMIGNAQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Assessment of dimensional stability of novel VPES impression material at different time intervals with standard disinfectants.: This research evaluates the dimensional stability of a novel vinyl polyether silicone (VPES) impression material when exposed to various disinfectants, including glutaraldehyde solutions, demonstrating its reliability for dental applications (Ahuja et al., 2024).

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Irene Costantini et al.
Scientific reports, 5, 9808-9808 (2015-05-08)
Extensive mapping of neuronal connections in the central nervous system requires high-throughput µm-scale imaging of large volumes. In recent years, different approaches have been developed to overcome the limitations due to tissue light scattering. These methods are generally developed to
Stéphane Jauréguiberry et al.
Blood, 124(2), 167-175 (2014-05-27)
Patients with severe malaria treated with artesunate sometimes experience a delayed hemolytic episode. Artesunate (AS) induces pitting, a splenic process whereby dead parasites are expelled from their host erythrocytes. These once-infected erythrocytes then return to the circulation. We analyzed hematologic
Andri K Riau et al.
Investigative ophthalmology & visual science, 55(5), 3186-3194 (2014-04-26)
To compare flap adhesion strength, stromal bed quality, and tissue responses after flap preparation using nJ- and μJ-energy level femtosecond lasers. All corneal flaps were created by either VisuMax laser (μJ-energy level) or femto-LDV (nJ-energy level). Flap adhesion strength in
Stephanie J Bissel et al.
Journal of neuropathology and experimental neurology, 74(8), 767-777 (2015-06-27)
Human parechovirus 3 (HPeV3) is a picornavirus associated with neurologic disease in neonates. Human parechovirus 3 infection of preterm and term infants is associated with seizures and destructive periventricular white matter lesions. Despite unremarkable cerebrospinal fluid (CSF), HPeV3 RNA can
Jamie Hinks et al.
Applied and environmental microbiology, 81(6), 1949-1958 (2015-01-13)
The modification of microbial membranes to achieve biotechnological strain improvement with exogenous small molecules, such as oligopolyphenylenevinylene-conjugated oligoelectrolyte (OPV-COE) membrane insertion molecules (MIMs), is an emerging biotechnological field. Little is known about the interactions of OPV-COEs with their target, the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service