Skip to Content
Merck
All Photos(1)

Key Documents

94681

Supelco

Methyl 3,5-dinitrobenzoate

Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

3,5-Dinitrobenzoic acid methyl ester, NSC 7317

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(O2N)2C6H3CO2CH3
CAS Number:
Molecular Weight:
226.14
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

Standard for quantitative NMR
TraceCERT®

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

qNMR: suitable

mp

107-109 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
pharmaceutical

SMILES string

COC(=O)c1cc(cc(c1)[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C8H6N2O6/c1-16-8(11)5-2-6(9(12)13)4-7(3-5)10(14)15/h2-4H,1H3

InChI key

POGCCFLNFPIIGW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Check out our entire range of quantitative NMR standards (qNMR standards)

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Method development in quantitative NMR towards metrologically traceable organic certified reference materials used as 31P qNMR standards.
Weber, M., Hellriegel, C., Rueck, A., Wuethrich, J., Jenks, P., and Obkircher, M.
Analytical and Bioanalytical Chemistry (2014)
Using high-performance quantitative NMR (HP-qNMR) for certifying traceable and highly accurate purity values of organic reference materials with uncertainties <0.1 %
Weber, M., Hellriegel, C., Rueck, A., Sauermoser, R., and Wuethrich, J.
Accreditation and Quality Assurance, 18, 91-98 (2013)
Michael Weber et al.
Journal of pharmaceutical and biomedical analysis, 93, 102-110 (2013-11-05)
Quantitative NMR spectroscopy (qNMR) is gaining interest across both analytical and industrial research applications and has become an essential tool for the content assignment and quantitative determination of impurities. The key benefits of using qNMR as measurement method for the

Articles

NMR shift charts assist in identifying impurities in deuterated solvents, aiding accurate chemical analysis.

NMR shift charts assist in identifying impurities in deuterated solvents, aiding accurate chemical analysis.

NMR shift charts assist in identifying impurities in deuterated solvents, aiding accurate chemical analysis.

NMR shift charts assist in identifying impurities in deuterated solvents, aiding accurate chemical analysis.

See All

Related Content

ChemisTwin automates NMR result interpretation, saving time with structure confirmation and quantitative NMR measurements using electronic Reference Materials.

ChemisTwin automates NMR result interpretation, saving time with structure confirmation and quantitative NMR measurements using electronic Reference Materials.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service