Skip to Content
Merck
All Photos(1)

Key Documents

900518

Sigma-Aldrich

Trifluoroacetic acid

anhydrous, ZerO2®, ≥99.0%

Synonym(s):

Degassed and low oxygen trifluoroacetic acid, Trifluoroacetic acid ZerO2®, TFA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3COOH
CAS Number:
Molecular Weight:
114.02
Beilstein:
742035
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.21

grade

ZerO2®

Quality Level

vapor density

3.9 (vs air)

vapor pressure

97.5 mmHg ( 20 °C)

Assay

≥99.0%

form

liquid

quality

anhydrous

refractive index

n20/D 1.3 (lit.)

bp

72.4 °C (lit.)

mp

−15.4 °C (lit.)

density

1.489 g/mL at 20 °C (lit.)

SMILES string

OC(C(F)(F)F)=O

InChI

1S/C2HF3O2/c3-2(4,5)1(6)7/h(H,6,7)

InChI key

DTQVDTLACAAQTR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

ZerO2 products are rigorously degassed with highly pure inert gas providing solvents and solutions (anhydrous if specified) with very low residual oxygen content.

Legal Information

ZerO2 is a registered trademark of Sigma-Aldrich Co. LLC

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

>212.0 °F - Pensky-Martens closed cup

Flash Point(C)

> 100 °C - Pensky-Martens closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service