Skip to Content
Merck
All Photos(1)

Key Documents

87997

Supelco

Tetrapentylammonium bromide

suitable for ion pair chromatography, LiChropur, ≥99.0% (AT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)4]4N(Br)
CAS Number:
Molecular Weight:
378.47
Beilstein:
3658171
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NB.21

description

cationic

Quality Level

Assay

≥99.0% (AT)

form

powder

quality

LiChropur

technique(s)

ion pair chromatography: suitable

mp

100-101 °C (lit.)

λ

10 % in methanol

UV absorption

λ: 240 nm Amax: 0.04
λ: 250 nm Amax: 0.03
λ: 260 nm Amax: 0.02
λ: 500 nm Amax: 0.02

suitability

corresponds to standard for filter test

SMILES string

[Br-].CCCCC[N+](CCCCC)(CCCCC)CCCCC

InChI

1S/C20H44N.BrH/c1-5-9-13-17-21(18-14-10-6-2,19-15-11-7-3)20-16-12-8-4;/h5-20H2,1-4H3;1H/q+1;/p-1

InChI key

SPALIFXDWQTXKS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Tetrapentylammonium bromide is a quaternary ammonium bromide, with pentyl chains and a bromide counter-ion, which is widely used as an ion-pair reagent in analytical applications.

Application

Tetrapentylammonium bromide may be used as a standard, to examine the effects of annealing membranes formed by mixture-casting ”Nafion” mainly based on the selectivity, ion-exchange capacity, and mass transport in the membranes. It may also be used as an ion pair reagent in the analysis of antioxidant phenolic compounds in wines, juices using two-dimensional liquid chromatography and sulfonated lignins in agrochemical formulations using one-, two-dimensional liquid chromatography techniques.

Other Notes

Additive used in ion-pair chromatography; Induces longer retention times than does the tetrabutyl homologue

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comprehensive two-dimensional liquid chromatography in the analysis of antioxidant phenolic compounds in wines and juices
Kivilompolo M, et al.
Analytical and Bioanalytical Chemistry, 391(1), 373-380 (2008)
K. Sagara et al.
Journal of Chromatography A, 328, 289-289 (1985)
A.T. Hunter
Journal of Chromatography A, 319, 319-319 (1985)
One-dimensional and two-dimensional liquid chromatography of sulphonated lignins
Brudin S, et al.
Journal of Chromatography A, 1201(2), 196-201 (2008)
Effects of annealing on mixture-cast membranes of [TM="Nafion"] and quaternary ammonium bromide salts
Thomas JT, et al.
Journal of Membrane Science , 213 (1-2), 55-66 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service