Skip to Content
Merck
All Photos(1)

Key Documents

8.20894

Sigma-Aldrich

Nitromethane

for synthesis

Synonym(s):

Nitromethane, Mononitromethane, Nitrocarbol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH3NO2
CAS Number:
Molecular Weight:
61.04
MDL number:
UNSPSC Code:
12352102
EC Index Number:
200-876-6
NACRES:
NA.22
Assay:
≥98% (GC)
bp:
101.2 °C/1013 hPa
vapor pressure:
36.4 hPa ( 20 °C)
Pricing and availability is not currently available.

vapor pressure

36.4 hPa ( 20 °C)

Quality Level

Assay

≥98% (GC)

form

liquid

potency

940 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

expl. lim.

7.3-63.0 % (v/v)

pH

6.4 (20 °C, 0.6 g/L in H2O)

bp

101.2 °C/1013 hPa

mp

-29 °C

transition temp

flash point 35.6 °C

solubility

105 g/L

density

1.14 g/cm3 at 20 °C

functional group

nitro

storage temp.

15-25°C

SMILES string

[N+](=O)([O-])C

InChI

1S/CH3NO2/c1-2(3)4/h1H3

InChI key

LYGJENNIWJXYER-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98,0 Area %
Density (d 20 °C/ 4 °C): 1,134 - 1,138
Identity (IR): conforms

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Flam. Liq. 3 - Repr. 2

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 2

Flash Point(F)

95.0 °F - closed cup

Flash Point(C)

35 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service