Skip to Content
Merck
All Photos(1)

Key Documents

59964

Supelco

Histamine

analytical standard

Synonym(s):

2-(4-Imidazolyl)ethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9N3
CAS Number:
Molecular Weight:
111.15
Beilstein:
2012
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.0% (TLC)
≥97.0% (anhydrous basis, NT)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤4.0% water

color

white to light yellow

bp

167 °C/0.8 mmHg (lit.)

mp

83-84 °C (lit.)

format

neat

storage temp.

−20°C

SMILES string

NCCc1c[nH]cn1

InChI

1S/C5H9N3/c6-2-1-5-3-7-4-8-5/h3-4H,1-2,6H2,(H,7,8)

InChI key

NTYJJOPFIAHURM-UHFFFAOYSA-N

Gene Information

human ... HRH1(3269)

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Endogenous H1 and H2 histamine receptor agonist; H1 activation mobilizes Ca2+; H2 activation stimulates adenylate cyclase activity in neurons; activates nitric oxide synthetase; potent vasodilator.
Endogenous H1 and H2 histamine receptor agonist; activates nitric oxide synthetase; potent vasodilator.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P J Turner et al.
Pediatric allergy and immunology : official publication of the European Society of Pediatric Allergy and Immunology, 25(7), 657-661 (2014-10-10)
Most children with egg allergy tolerate egg in baked foods, such as cake, but tolerance cannot be predicted with conventional allergy testing. We hypothesized that the skin prick test (SPT) wheal to unprocessed raw egg might predict tolerance of baked
Ye Zhang et al.
Investigative ophthalmology & visual science, 56(1), 563-570 (2015-01-13)
We estimate and compare change in iris cross-sectional area (IA) after physiologic and pharmacologic mydriasis in subjects with different dominant mechanisms for primary angle closure. Anterior segment optical coherence tomography (AS-OCT) measurements in light, dark, and following pharmacologic dilation were
Andrea Strasser et al.
Trends in pharmacological sciences, 34(1), 13-32 (2012-12-12)
Histamine is a biogenic amine that exerts its biological effects as a neurotransmitter and local mediator via four histamine receptor (HR) subtypes (H(x)Rs) - H(1)R, H(2)R, H(3)R, and H(4)R - belonging to the superfamily of G-protein-coupled receptors (GPCRs). All four
Katsuya Nagai et al.
Amino acids, 43(1), 97-109 (2012-03-01)
L-carnosine (β-alanyl-L-histidine; CAR) is synthesized in mammalian skeletal muscle. Although the physiological roles of CAR have not yet been clarified, there is evidence that the release of CAR from skeletal muscle during physical exercise affects autonomic neurotransmission and physiological functions.
W Tang et al.
Oncogene, 33(20), 2629-2638 (2013-06-12)
Recent studies suggested that cancer stem cells (CSCs) are capable of differentiating into endothelial cells and tumor endothelium may be derived from CSCs. But the mechanism remains unclear. We showed that vascular endothelial growth factor (VEGF) induced the expression of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service