Skip to Content
Merck
All Photos(1)

Key Documents

43034

Supelco

Bisphenol S

analytical standard

Synonym(s):

4,4′-Sulfonyldiphenol, BPS, 4-Hydroxyphenyl sulfone, Bis(4-hydroxyphenyl) sulfone, Bisphenol S

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2S(C6H4OH)2
CAS Number:
Molecular Weight:
250.27
Beilstein:
2052954
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

245-250 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

Oc1ccc(cc1)S(=O)(=O)c2ccc(O)cc2

InChI

1S/C12H10O4S/c13-9-1-5-11(6-2-9)17(15,16)12-7-3-10(14)4-8-12/h1-8,13-14H

InChI key

VPWNQTHUCYMVMZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bisphenol S (BPS), belonging to the class of bisphenols, is a structural analog of bisphenol A (BPA), used in a variety of industrial applications and products marketed as BPA-free. It is commonly identified in food and beverages, environmental samples, biological matrices, etc. It is also reported to be used in the production of epoxy resins. BPS is also found to be stable compared to BPA at elevated temperature and sunlight conditions.
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Application

Bisphenol S may be used as an analytical reference standard for the determination of the analyte in:
  • Beverages using liquid chromatography coupled to tandem mass spectrometry (LC-MS/MS).
  • Biological samples using high performance liquid chromatography coupled to tandem mass spectrometry (HPLC-MS/MS).
  • Environmental water samples using high performance liquid chromatography (HPLC) with ultraviolet (UV) detection.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bisphenol S-and bisphenol A-induced adipogenesis of murine preadipocytes occurs through direct peroxisome proliferator-activated receptor gamma activation
Ahmed S and Atlas E
International journal of obesity (2005), 40(10), 1566-1573 (2016)
Vortex-assisted liquid-liquid microextraction of bisphenol S prior to its determination by HPLC with UV detection
Diao P-C, et al.
Microchimica Acta, 182(15-16), 2593-2600 (2015)
Analysis of bisphenols in soft drinks by on-line solid phase extraction fast liquid chromatography-tandem mass spectrometry.
Gallart-Ayala, et al.
Analytica Chimica Acta, 683(2), 227-233 (2011)
Tara R Catron et al.
Toxicological sciences : an official journal of the Society of Toxicology, 167(2), 468-483 (2018-10-16)
Host-associated microbiota can biotransform xenobiotics, mediate health effects of chemical exposure, and play important roles in early development. Bisphenol A (BPA) is a widespread environmental chemical that has been associated with adverse endocrine and neurodevelopmental effects, some of which may
Automated on-line column-switching high-performance liquid chromatography isotope dilution tandem mass spectrometry method for the quantification of bisphenol A, bisphenol F, bisphenol S, and 11 other phenols in urine
Zhou X, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 944, 152-156 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service