Skip to Content
Merck
All Photos(3)

Key Documents

410225

Sigma-Aldrich

Glycine

≥98.5%, ACS reagent for peptide synthesis

Synonym(s):

Aminoacetic acid, Aminoethanoic acid, Glycocoll

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2COOH
CAS Number:
Molecular Weight:
75.07
Beilstein:
635782
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.21

product name

Glycine, ACS reagent, ≥98.5%

grade

ACS reagent

Quality Level

Assay

≥98.5%

form

powder, crystals or granules

impurities

H2SO4, passes test (darkened)
Hydrolyzable subsances, passes test

ign. residue

≤0.1%

color

white

pKa (25 °C)

(1) 2.35, (2) 9.60
2.35

mp

240 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤0.005%
sulfate (SO42-): ≤0.005%

cation traces

NH4+: ≤0.005%
heavy metals: ≤0.002% (by ICP)

application(s)

peptide synthesis

SMILES string

NCC(O)=O

InChI

1S/C2H5NO2/c3-1-2(4)5/h1,3H2,(H,4,5)

InChI key

DHMQDGOQFOQNFH-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Glycine is the simplest amino acid building block.

Application

Glycine has been used in whole-cell patch clamp recording to measure inward NMDA (N-methyl-D-aspartate) currents. It has also been used as a model organic system in crystallography to demonstrate a novel acoustic nebulization platform.

Biochem/physiol Actions

Glycine is a non-essential amino acid. Influx of calcium through the cell membrane is mediated by glycine-gated channel. Glycine participates in the synthesis of porphyrins, purine and serine. It also serves as a competitive agonist for glutamate in binding to the NMDA (N-methyl-D-aspartate) receptors. Glycine synthesis might be increased in rapidly proliferating cancer cells, due to increased demand for the amino acid.
Inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Controversial Effects of D-Amino Acid Oxidase Activator (DAOA)/G72 on D-Amino Acid Oxidase (DAO) Activity in Human Neuronal, Astrocyte and Kidney Cell Lines: The N-methyl D-aspartate (NMDA) Receptor Hypofunction Point of View
Vinita Jagannath
Frontiers in Molecular Neuroscience (2017)
A Novel Acoustomicrofluidic Nebulization Technique Yielding New Crystallization Morphologies
Heba Ahmed
Advanced Materials, 30(3) (2018)
Amino acids: metabolism, functions, and nutrition
Guoyao Wu
Amino Acids, 37(1), 1-17 (2009)
Metabolite Profiling Identifies a Key Role for Glycine in Rapid Cancer Cell Proliferation
Mohit Jain
Science, 336(6084), 1040-1044 (2012)
Alexander Carvajal-González et al.
Brain : a journal of neurology, 137(Pt 8), 2178-2192 (2014-06-22)
The clinical associations of glycine receptor antibodies have not yet been described fully. We identified prospectively 52 antibody-positive patients and collated their clinical features, investigations and immunotherapy responses. Serum glycine receptor antibody endpoint titres ranged from 1:20 to 1:60 000.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service